Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41076146-41076319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOP1
Entrez Gene ID
7150
Gene Name
topoisomerase (DNA) I
Gene Aliases
TOPI
UniGene
Hs.472737
Gene Chromosome Location
Chr.20: 41028822-41124487 on Build GRCh38
Transcripts
NM_003286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6930638 c.237C>G p.D79E Chr.20: 41076252-41076252 on Build GRCh38
5662828 c.196A>T p.K66* Chr.20: 41076211-41076211 on Build GRCh38
4967854 c.271G>C p.E91Q Chr.20: 41076286-41076286 on Build GRCh38
723682 c.158A>G p.E53G Chr.20: 41076173-41076173 on Build GRCh38
723681 c.183C>G p.H61Q Chr.20: 41076198-41076198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762757738 Chr.20: 41076185-41076185 on Build GRCh38
rs773155637 Chr.20: 41076174-41076174 on Build GRCh38
rs911983921 Chr.20: 41076259-41076264 on Build GRCh38
rs751566633 Chr.20: 41076221-41076221 on Build GRCh38
rs953420719 Chr.20: 41076255-41076255 on Build GRCh38
rs61756255 Chr.20: 41076256-41076256 on Build GRCh38
rs145530164 Chr.20: 41076285-41076285 on Build GRCh38
rs755282743 Chr.20: 41076222-41076222 on Build GRCh38
rs767844946 Chr.20: 41076234-41076234 on Build GRCh38
rs778237366 Chr.20: 41076281-41076281 on Build GRCh38
rs777893669 Chr.20: 41076255-41076266 on Build GRCh38
rs766516532 Chr.20: 41076186-41076186 on Build GRCh38
rs752903786 Chr.20: 41076243-41076243 on Build GRCh38
rs889429048 Chr.20: 41076252-41076252 on Build GRCh38

View Full Product Details