Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43102522-43102648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RET
Entrez Gene ID
5979
Gene Name
ret proto-oncogene
Gene Aliases
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1, RET51
UniGene
Hs.350321
Gene Chromosome Location
Chr.10: 43077069-43130351 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836876 c.518C>T p.S173F Chr.10: 43102522-43102522 on Build GRCh38
5359844 c.530G>A p.R177Q Chr.10: 43102534-43102534 on Build GRCh38
5077233 c.529C>T p.R177W Chr.10: 43102533-43102533 on Build GRCh38
4014152 c.613A>G p.R205G Chr.10: 43102617-43102617 on Build GRCh38
6969026 c.542C>A p.P181H Chr.10: 43102546-43102546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048022444 Chr.10: 43102557-43102557 on Build GRCh38
rs764162555 Chr.10: 43102565-43102565 on Build GRCh38
rs898525501 Chr.10: 43102606-43102606 on Build GRCh38
rs764910706 Chr.10: 43102595-43102595 on Build GRCh38
rs755007607 Chr.10: 43102611-43102611 on Build GRCh38
rs76449634 Chr.10: 43102542-43102542 on Build GRCh38
rs1006106202 Chr.10: 43102600-43102600 on Build GRCh38
rs775086466 Chr.10: 43102548-43102548 on Build GRCh38
rs750675926 Chr.10: 43102597-43102597 on Build GRCh38
rs753301491 Chr.10: 43102554-43102554 on Build GRCh38
rs1008123818 Chr.10: 43102581-43102581 on Build GRCh38
rs751572082 Chr.10: 43102608-43102608 on Build GRCh38
rs144801580 Chr.10: 43102552-43102552 on Build GRCh38
rs55810667 Chr.10: 43102601-43102601 on Build GRCh38
rs368116579 Chr.10: 43102586-43102586 on Build GRCh38
rs781750106 Chr.10: 43102628-43102628 on Build GRCh38
rs753707182 Chr.10: 43102570-43102570 on Build GRCh38
rs370736139 Chr.10: 43102543-43102543 on Build GRCh38
rs780120451 Chr.10: 43102607-43102607 on Build GRCh38
rs886046986 Chr.10: 43102600-43102600 on Build GRCh38
rs76736111 Chr.10: 43102596-43102596 on Build GRCh38

View Full Product Details