Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102324557-102324706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC3
Entrez Gene ID
330
Gene Name
baculoviral IAP repeat containing 3
Gene Aliases
AIP1, API2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49, c-IAP2
UniGene
Hs.127799
Gene Chromosome Location
Chr.11: 102317450-102339403 on Build GRCh38
Transcripts
NM_001165.4 NM_182962.2 XM_017017643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2108117 c.49G>A p.A17T Chr.11: 102324558-102324558 on Build GRCh38
4990037 c.71A>T p.Y24F Chr.11: 102324580-102324580 on Build GRCh38
1704480 c.118C>T p.P40S Chr.11: 102324627-102324627 on Build GRCh38
1746003 c.73G>T p.D25Y Chr.11: 102324582-102324582 on Build GRCh38
367076 c.92A>C p.Y31S Chr.11: 102324601-102324601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770856613 Chr.11: 102324581-102324581 on Build GRCh38
rs1055078 Chr.11: 102324638-102324638 on Build GRCh38
rs761068492 Chr.11: 102324591-102324591 on Build GRCh38
rs1055079 Chr.11: 102324665-102324665 on Build GRCh38
rs190140643 Chr.11: 102324614-102324614 on Build GRCh38
rs370738985 Chr.11: 102324582-102324582 on Build GRCh38
rs762397304 Chr.11: 102324664-102324664 on Build GRCh38
rs978016131 Chr.11: 102324657-102324657 on Build GRCh38
rs1035350946 Chr.11: 102324612-102324612 on Build GRCh38
rs775814824 Chr.11: 102324588-102324588 on Build GRCh38
rs759088779 Chr.11: 102324681-102324681 on Build GRCh38
rs375640711 Chr.11: 102324587-102324587 on Build GRCh38
rs765765029 Chr.11: 102324676-102324676 on Build GRCh38
rs751101959 Chr.11: 102324677-102324677 on Build GRCh38
rs777182261 Chr.11: 102324630-102324630 on Build GRCh38

View Full Product Details