Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119273825-119273990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBL
Entrez Gene ID
867
Gene Name
Cbl proto-oncogene
Gene Aliases
C-CBL, CBL2, FRA11B, NSLL, RNF55
UniGene
Hs.504096
Gene Chromosome Location
Chr.11: 119206276-119308149 on Build GRCh38
Transcripts
NM_005188.3 XM_011543057.2 XM_017018511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341123 c.647G>C p.S216T Chr.11: 119273924-119273924 on Build GRCh38
6947186 c.652G>A p.G218R Chr.11: 119273929-119273929 on Build GRCh38
343953 c.650C>A p.S217Y Chr.11: 119273927-119273927 on Build GRCh38
248733 c.592A>C p.T198P Chr.11: 119273869-119273869 on Build GRCh38
6976217 c.591-2A>T p.? Chr.11: 119273866-119273866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895664191 Chr.11: 119273933-119273933 on Build GRCh38
rs869155403 Chr.11: 119273940-119273940 on Build GRCh38
rs34832489 Chr.11: 119273939-119273939 on Build GRCh38
rs769731615 Chr.11: 119273854-119273855 on Build GRCh38
rs767162260 Chr.11: 119273902-119273902 on Build GRCh38
rs1052915853 Chr.11: 119273932-119273932 on Build GRCh38
rs763760976 Chr.11: 119273923-119273923 on Build GRCh38
rs926023115 Chr.11: 119273878-119273878 on Build GRCh38
rs759082853 Chr.11: 119273888-119273888 on Build GRCh38
rs760192666 Chr.11: 119273920-119273920 on Build GRCh38
rs397517082 Chr.11: 119273872-119273872 on Build GRCh38
rs949933368 Chr.11: 119273937-119273937 on Build GRCh38
rs747041808 Chr.11: 119273859-119273859 on Build GRCh38
rs773611782 Chr.11: 119273941-119273941 on Build GRCh38
rs1057518053 Chr.11: 119273942-119273942 on Build GRCh38
rs775309469 Chr.11: 119273911-119273911 on Build GRCh38
rs749291716 Chr.11: 119273860-119273860 on Build GRCh38
rs1044358721 Chr.11: 119273947-119273947 on Build GRCh38
rs757950499 Chr.11: 119273901-119273901 on Build GRCh38

View Full Product Details