Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121483685-121483854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
UniGene
Hs.533683
Gene Chromosome Location
Chr.10: 121478330-121598656 on Build GRCh38
Transcripts
NM_000141.4 NM_001144913.1 NM_001144914.1 NM_001144915.1 NM_001144916.1 NM_001144917.1 NM_001144918.1 NM_001144919.1 NM_001320654.1 NM_001320658.1 NM_022970.3 NM_023029.2 NR_073009.1 XM_006717708.2 XM_006717710.3 XM_006717711.2 XM_006717712.2 XM_017015920.1 XM_017015921.1 XM_017015922.1 XM_017015923.1 XM_017015924.1 XM_017015925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5477129 c.2284C>A p.L762I Chr.10: 121483718-121483718 on Build GRCh38
6938993 c.2290C>G p.L764V Chr.10: 121483712-121483712 on Build GRCh38
3434721 c.2235C>T p.S745S Chr.10: 121483764-121483764 on Build GRCh38
6941577 c.2279G>A p.R760Q Chr.10: 121483723-121483723 on Build GRCh38
3934957 c.2299G>A p.E767K Chr.10: 121483700-121483700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116895810 Chr.10: 121483761-121483761 on Build GRCh38
rs142639988 Chr.10: 121483767-121483767 on Build GRCh38
rs781182356 Chr.10: 121483749-121483749 on Build GRCh38
rs751731823 Chr.10: 121483739-121483739 on Build GRCh38
rs745947630 Chr.10: 121483753-121483753 on Build GRCh38
rs912905439 Chr.10: 121483718-121483718 on Build GRCh38
rs754928713 Chr.10: 121483754-121483754 on Build GRCh38
rs55774317 Chr.10: 121483723-121483723 on Build GRCh38
rs777950855 Chr.10: 121483738-121483738 on Build GRCh38
rs868056281 Chr.10: 121483724-121483724 on Build GRCh38
rs753103160 Chr.10: 121483716-121483716 on Build GRCh38
rs925882164 Chr.10: 121483828-121483828 on Build GRCh38
rs375719482 Chr.10: 121483768-121483768 on Build GRCh38
rs757501816 Chr.10: 121483748-121483748 on Build GRCh38
rs746597657 Chr.10: 121483823-121483823 on Build GRCh38
rs894067673 Chr.10: 121483806-121483806 on Build GRCh38
rs41294229 Chr.10: 121483710-121483710 on Build GRCh38
rs773160247 Chr.10: 121483819-121483819 on Build GRCh38
rs747941020 Chr.10: 121483769-121483769 on Build GRCh38
rs771807547 Chr.10: 121483770-121483770 on Build GRCh38

View Full Product Details