Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61683377-61683515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRIP1
Entrez Gene ID
83990
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Gene Aliases
BACH1, FANCJ, OF
UniGene
Hs.128903
Gene Chromosome Location
Chr.17: 61679186-61864120 on Build GRCh38
Transcripts
NM_032043.2 XM_011525332.2 XM_011525333.2 XM_011525334.2 XM_011525335.2 XM_011525336.2 XM_011525337.2 XM_011525338.2 XM_017025200.1 XM_017025201.1 XM_017025202.1 XM_017025203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5575450 c.3622G>T p.D1208Y Chr.17: 61683424-61683424 on Build GRCh38
6922877 c.3617T>G p.I1206S Chr.17: 61683429-61683429 on Build GRCh38
1384992 c.3623A>G p.D1208G Chr.17: 61683423-61683423 on Build GRCh38
5659975 c.3664G>T p.E1222* Chr.17: 61683382-61683382 on Build GRCh38
5575537 c.3590G>T p.G1197V Chr.17: 61683456-61683456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764848326 Chr.17: 61683479-61683479 on Build GRCh38
rs941141257 Chr.17: 61683434-61683434 on Build GRCh38
rs763298204 Chr.17: 61683489-61683489 on Build GRCh38
rs760589795 Chr.17: 61683424-61683424 on Build GRCh38
rs776010326 Chr.17: 61683441-61683441 on Build GRCh38
rs548674096 Chr.17: 61683419-61683419 on Build GRCh38
rs367610893 Chr.17: 61683487-61683487 on Build GRCh38
rs768156067 Chr.17: 61683431-61683431 on Build GRCh38
rs139539831 Chr.17: 61683426-61683426 on Build GRCh38
rs786201962 Chr.17: 61683484-61683484 on Build GRCh38
rs112214651 Chr.17: 61683417-61683417 on Build GRCh38
rs730881628 Chr.17: 61683447-61683447 on Build GRCh38
rs876659839 Chr.17: 61683451-61683451 on Build GRCh38
rs766709841 Chr.17: 61683490-61683490 on Build GRCh38
rs587782615 Chr.17: 61683438-61683439 on Build GRCh38
rs374392860 Chr.17: 61683410-61683410 on Build GRCh38
rs587781677 Chr.17: 61683456-61683456 on Build GRCh38
rs761405340 Chr.17: 61683475-61683475 on Build GRCh38

View Full Product Details