Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39230192-39230351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFB
Entrez Gene ID
5155
Gene Name
platelet derived growth factor subunit B
Gene Aliases
IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis
UniGene
Hs.1976
Gene Chromosome Location
Chr.22: 39223359-39245055 on Build GRCh38
Transcripts
NM_002608.3 NM_033016.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399539 c.457-1G>A p.? Chr.22: 39230229-39230229 on Build GRCh38
5393635 c.477G>A p.V159V Chr.22: 39230208-39230208 on Build GRCh38
4713286 c.413T>G p.V138G Chr.22: 39230227-39230227 on Build GRCh38
4388422 c.478C>T p.R160W Chr.22: 39230207-39230207 on Build GRCh38
1308170 c.423C>T p.I141I Chr.22: 39230217-39230217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048321530 Chr.22: 39230248-39230248 on Build GRCh38
rs977419415 Chr.22: 39230330-39230330 on Build GRCh38
rs1019299647 Chr.22: 39230283-39230283 on Build GRCh38
rs754421060 Chr.22: 39230274-39230274 on Build GRCh38
rs879090808 Chr.22: 39230296-39230296 on Build GRCh38
rs764345359 Chr.22: 39230250-39230250 on Build GRCh38
rs201990837 Chr.22: 39230216-39230216 on Build GRCh38
rs945973570 Chr.22: 39230294-39230294 on Build GRCh38
rs1029570235 Chr.22: 39230308-39230308 on Build GRCh38
rs900922242 Chr.22: 39230291-39230291 on Build GRCh38
rs922409591 Chr.22: 39230285-39230285 on Build GRCh38
rs774559047 Chr.22: 39230217-39230217 on Build GRCh38
rs199785704 Chr.22: 39230257-39230257 on Build GRCh38
rs931227495 Chr.22: 39230232-39230232 on Build GRCh38
rs369782393 Chr.22: 39230241-39230241 on Build GRCh38
rs1021895713 Chr.22: 39230319-39230319 on Build GRCh38
rs187298550 Chr.22: 39230282-39230282 on Build GRCh38
rs886762911 Chr.22: 39230281-39230281 on Build GRCh38
rs928703060 Chr.22: 39230332-39230332 on Build GRCh38
rs535004629 Chr.22: 39230301-39230301 on Build GRCh38
rs779458038 Chr.22: 39230271-39230271 on Build GRCh38
rs754270223 Chr.22: 39230251-39230251 on Build GRCh38
rs967346718 Chr.22: 39230320-39230320 on Build GRCh38
rs757757910 Chr.22: 39230267-39230267 on Build GRCh38
rs953612861 Chr.22: 39230313-39230313 on Build GRCh38
rs767707710 Chr.22: 39230223-39230223 on Build GRCh38
rs990385889 Chr.22: 39230314-39230314 on Build GRCh38

View Full Product Details