Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193135526-193135678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_024529.4 XM_006711537.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441868 c.415C>G p.R139G Chr.1: 193135581-193135581 on Build GRCh38
4670742 c.383C>T p.A128V Chr.1: 193135549-193135549 on Build GRCh38
330598 c.415C>T p.R139* Chr.1: 193135581-193135581 on Build GRCh38
6939210 c.423+1G>A p.? Chr.1: 193135590-193135590 on Build GRCh38
5899810 c.413C>T p.P138L Chr.1: 193135579-193135579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369542555 Chr.1: 193135578-193135578 on Build GRCh38
rs748367546 Chr.1: 193135618-193135618 on Build GRCh38
rs773739963 Chr.1: 193135608-193135613 on Build GRCh38
rs774763324 Chr.1: 193135615-193135615 on Build GRCh38
rs976186333 Chr.1: 193135629-193135629 on Build GRCh38
rs908205344 Chr.1: 193135624-193135624 on Build GRCh38
rs770561055 Chr.1: 193135600-193135601 on Build GRCh38
rs765308159 Chr.1: 193135647-193135647 on Build GRCh38
rs776500275 Chr.1: 193135628-193135628 on Build GRCh38
rs547890731 Chr.1: 193135648-193135648 on Build GRCh38
rs768821497 Chr.1: 193135604-193135604 on Build GRCh38
rs769052041 Chr.1: 193135557-193135557 on Build GRCh38
rs375460045 Chr.1: 193135619-193135619 on Build GRCh38
rs745334057 Chr.1: 193135625-193135627 on Build GRCh38
rs923317075 Chr.1: 193135650-193135650 on Build GRCh38
rs780017801 Chr.1: 193135560-193135560 on Build GRCh38
rs749344710 Chr.1: 193135595-193135595 on Build GRCh38

View Full Product Details