Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18742948-18743118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC1
Entrez Gene ID
23373
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
MECT1, TORC-1, TORC1, WAMTP1
UniGene
Hs.371096
Gene Chromosome Location
Chr.19: 18683615-18782333 on Build GRCh38
Transcripts
NM_001098482.1 NM_015321.2 XM_005259833.3 XM_005259834.1 XM_005259835.3 XM_005259836.3 XM_006722710.3 XM_011527842.2 XM_017026536.1 XM_017026537.1 XM_017026538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835321 c.214G>A p.G72R Chr.19: 18742997-18742997 on Build GRCh38
6294744 c.173G>T p.G58V Chr.19: 18742956-18742956 on Build GRCh38
4834677 c.215G>A p.G72E Chr.19: 18742998-18742998 on Build GRCh38
1391716 c.224C>T p.T75I Chr.19: 18743007-18743007 on Build GRCh38
6294743 c.173G>T p.G58V Chr.19: 18742956-18742956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754533421 Chr.19: 18743002-18743002 on Build GRCh38
rs755728185 Chr.19: 18742984-18742984 on Build GRCh38
rs774222165 Chr.19: 18743057-18743057 on Build GRCh38
rs754013603 Chr.19: 18743062-18743062 on Build GRCh38
rs757003640 Chr.19: 18742974-18742974 on Build GRCh38
rs541696446 Chr.19: 18742969-18742969 on Build GRCh38
rs772517937 Chr.19: 18743017-18743017 on Build GRCh38
rs751456753 Chr.19: 18743061-18743061 on Build GRCh38
rs750264099 Chr.19: 18742976-18742976 on Build GRCh38
rs776924441 Chr.19: 18743038-18743038 on Build GRCh38
rs779223908 Chr.19: 18743008-18743008 on Build GRCh38
rs771044924 Chr.19: 18743037-18743037 on Build GRCh38
rs371207750 Chr.19: 18743048-18743048 on Build GRCh38
rs748737417 Chr.19: 18742998-18742998 on Build GRCh38
rs751500321 Chr.19: 18742973-18742973 on Build GRCh38
rs779827394 Chr.19: 18742996-18742996 on Build GRCh38
rs748698668 Chr.19: 18743015-18743015 on Build GRCh38
rs773690296 Chr.19: 18743019-18743019 on Build GRCh38
rs890922452 Chr.19: 18743047-18743047 on Build GRCh38
rs747419096 Chr.19: 18743029-18743029 on Build GRCh38
rs560753245 Chr.19: 18742975-18742975 on Build GRCh38

View Full Product Details