Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171391249-171391373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPM1
Entrez Gene ID
4869
Gene Name
nucleophosmin
Gene Aliases
B23, NPM
UniGene
Hs.557550
Gene Chromosome Location
Chr.5: 171387648-171410900 on Build GRCh38
Transcripts
NM_001037738.2 NM_002520.6 NM_199185.3 XM_005265920.3 XM_011534564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6279337 c.168G>A p.E56E Chr.5: 171391334-171391334 on Build GRCh38
302770 c.153T>G p.A51A Chr.5: 171391319-171391319 on Build GRCh38
6960116 c.176T>C p.I59T Chr.5: 171391342-171391342 on Build GRCh38
6470192 c.174C>T p.H58H Chr.5: 171391340-171391340 on Build GRCh38
6170651 c.157G>T p.A53S Chr.5: 171391323-171391323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003292380 Chr.5: 171391341-171391341 on Build GRCh38
rs765199192 Chr.5: 171391287-171391287 on Build GRCh38
rs142922469 Chr.5: 171391284-171391284 on Build GRCh38
rs939850854 Chr.5: 171391288-171391288 on Build GRCh38
rs755281834 Chr.5: 171391285-171391285 on Build GRCh38
rs908483380 Chr.5: 171391287-171391287 on Build GRCh38
rs894861205 Chr.5: 171391349-171391349 on Build GRCh38
rs758536527 Chr.5: 171391287-171391290 on Build GRCh38
rs752582321 Chr.5: 171391286-171391287 on Build GRCh38
rs774730997 Chr.5: 171391324-171391324 on Build GRCh38
rs778330876 Chr.5: 171391295-171391295 on Build GRCh38
rs749809103 Chr.5: 171391300-171391300 on Build GRCh38
rs751818162 Chr.5: 171391339-171391340 on Build GRCh38
rs777646492 Chr.5: 171391337-171391338 on Build GRCh38
rs757476342 Chr.5: 171391341-171391342 on Build GRCh38
rs759143092 Chr.5: 171391335-171391335 on Build GRCh38
rs199559300 Chr.5: 171391294-171391294 on Build GRCh38
rs771256760 Chr.5: 171391307-171391307 on Build GRCh38
rs759323042 Chr.5: 171391280-171391282 on Build GRCh38
rs142102985 Chr.5: 171391343-171391343 on Build GRCh38
rs781081167 Chr.5: 171391292-171391292 on Build GRCh38
rs997564714 Chr.5: 171391296-171391296 on Build GRCh38
rs748272368 Chr.5: 171391293-171391293 on Build GRCh38
rs372208958 Chr.5: 171391321-171391321 on Build GRCh38

View Full Product Details