Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52402173-52402324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6966297 c.2189G>T p.*730L Chr.3: 52402289-52402289 on Build GRCh38
6939917 c.2167_2174delCCCTACAA p.P723fs*>5 Chr.3: 52402304-52402311 on Build GRCh38
7335243 c.2185C>T p.Q729* Chr.3: 52402293-52402293 on Build GRCh38
7332103 c.2189G>A p.*730* Chr.3: 52402289-52402289 on Build GRCh38
422744 c.2188T>G p.*730G Chr.3: 52402290-52402290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926178223 Chr.3: 52402231-52402231 on Build GRCh38
rs771713346 Chr.3: 52402289-52402289 on Build GRCh38
rs748022925 Chr.3: 52402285-52402285 on Build GRCh38
rs748962519 Chr.3: 52402262-52402262 on Build GRCh38
rs139307137 Chr.3: 52402251-52402251 on Build GRCh38
rs773230722 Chr.3: 52402295-52402295 on Build GRCh38
rs1025761013 Chr.3: 52402215-52402215 on Build GRCh38
rs764618711 Chr.3: 52402241-52402241 on Build GRCh38
rs375252451 Chr.3: 52402252-52402252 on Build GRCh38
rs56898787 Chr.3: 52402243-52402243 on Build GRCh38
rs778397771 Chr.3: 52402269-52402269 on Build GRCh38
rs768488999 Chr.3: 52402264-52402264 on Build GRCh38
rs779640381 Chr.3: 52402256-52402256 on Build GRCh38
rs564062251 Chr.3: 52402228-52402228 on Build GRCh38
rs760537008 Chr.3: 52402303-52402303 on Build GRCh38

View Full Product Details