Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41128911-41129065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP9X
Entrez Gene ID
8239
Gene Name
ubiquitin specific peptidase 9, X-linked
Gene Aliases
DFFRX, FAF, FAM, MRX99, MRXS99F
UniGene
Hs.77578
Gene Chromosome Location
Chr.X: 41085420-41236579 on Build GRCh38
Transcripts
NM_001039590.2 NM_001039591.2 XM_005272675.4 XM_005272676.4

SNP Detail

SNP ID Chromosome Location
rs73480411 Chr.X: 41128935-41128935 on Build GRCh38
rs766015161 Chr.X: 41129020-41129020 on Build GRCh38
rs762490407 Chr.X: 41129017-41129017 on Build GRCh38
rs755870817 Chr.X: 41129031-41129031 on Build GRCh38
rs776650027 Chr.X: 41128968-41128968 on Build GRCh38
rs772817266 Chr.X: 41129007-41129007 on Build GRCh38
rs768572380 Chr.X: 41128959-41128959 on Build GRCh38
rs746775778 Chr.X: 41128956-41128956 on Build GRCh38
rs781534884 Chr.X: 41128947-41128947 on Build GRCh38
rs746092012 Chr.X: 41128977-41128977 on Build GRCh38
rs149866601 Chr.X: 41129034-41129034 on Build GRCh38
rs906242818 Chr.X: 41128957-41128957 on Build GRCh38

View Full Product Details