Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66389418-66389591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH5
Entrez Gene ID
1003
Gene Name
cadherin 5
Gene Aliases
7B4, CD144
UniGene
Hs.76206
Gene Chromosome Location
Chr.16: 66366607-66404786 on Build GRCh38
Transcripts
NM_001795.4 XM_011522801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1709261 c.706C>T p.R236W Chr.16: 66389447-66389447 on Build GRCh38
1709262 c.709G>A p.G237R Chr.16: 66389450-66389450 on Build GRCh38
5028146 c.691G>C p.D231H Chr.16: 66389432-66389432 on Build GRCh38
4802391 c.716C>A p.S239* Chr.16: 66389457-66389457 on Build GRCh38
6366360 c.707G>A p.R236Q Chr.16: 66389448-66389448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773614809 Chr.16: 66389453-66389453 on Build GRCh38
rs370349071 Chr.16: 66389566-66389566 on Build GRCh38
rs978846006 Chr.16: 66389563-66389563 on Build GRCh38
rs772333677 Chr.16: 66389554-66389554 on Build GRCh38
rs147201423 Chr.16: 66389464-66389464 on Build GRCh38
rs373529370 Chr.16: 66389568-66389568 on Build GRCh38
rs202178871 Chr.16: 66389481-66389481 on Build GRCh38
rs748519365 Chr.16: 66389466-66389468 on Build GRCh38
rs763466378 Chr.16: 66389471-66389471 on Build GRCh38
rs372943076 Chr.16: 66389517-66389517 on Build GRCh38
rs139763470 Chr.16: 66389457-66389457 on Build GRCh38
rs749874514 Chr.16: 66389440-66389440 on Build GRCh38
rs149810324 Chr.16: 66389458-66389458 on Build GRCh38
rs543649916 Chr.16: 66389463-66389463 on Build GRCh38
rs907777703 Chr.16: 66389503-66389503 on Build GRCh38
rs559066873 Chr.16: 66389551-66389551 on Build GRCh38
rs780802038 Chr.16: 66389484-66389484 on Build GRCh38
rs767815644 Chr.16: 66389450-66389450 on Build GRCh38
rs545781173 Chr.16: 66389532-66389532 on Build GRCh38
rs199524490 Chr.16: 66389448-66389448 on Build GRCh38
rs375908441 Chr.16: 66389541-66389541 on Build GRCh38
rs151052156 Chr.16: 66389443-66389443 on Build GRCh38
rs773561465 Chr.16: 66389556-66389556 on Build GRCh38
rs780472148 Chr.16: 66389536-66389536 on Build GRCh38
rs760953455 Chr.16: 66389562-66389562 on Build GRCh38
rs769625369 Chr.16: 66389551-66389551 on Build GRCh38
rs775037310 Chr.16: 66389552-66389552 on Build GRCh38
rs554877512 Chr.16: 66389447-66389447 on Build GRCh38
rs758374402 Chr.16: 66389470-66389470 on Build GRCh38

View Full Product Details