Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40176533-40176704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B
Entrez Gene ID
701
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Gene Aliases
BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1
UniGene
Hs.513645
Gene Chromosome Location
Chr.15: 40161009-40221136 on Build GRCh38
Transcripts
NM_001211.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6482494 c.524A>T p.E175V Chr.15: 40176574-40176574 on Build GRCh38
6871131 c.504C>T p.F168F Chr.15: 40176554-40176554 on Build GRCh38
6482493 c.482A>T p.E161V Chr.15: 40176574-40176574 on Build GRCh38
6871130 c.462C>T p.F154F Chr.15: 40176554-40176554 on Build GRCh38
3500793 c.574C>T p.Q192* Chr.15: 40176666-40176666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746666250 Chr.15: 40176661-40176661 on Build GRCh38
rs1050594239 Chr.15: 40176567-40176567 on Build GRCh38
rs546538543 Chr.15: 40176558-40176558 on Build GRCh38
rs764923350 Chr.15: 40176556-40176556 on Build GRCh38
rs532319590 Chr.15: 40176600-40176600 on Build GRCh38
rs756702953 Chr.15: 40176609-40176609 on Build GRCh38
rs552380700 Chr.15: 40176610-40176610 on Build GRCh38
rs749701720 Chr.15: 40176636-40176636 on Build GRCh38
rs761819521 Chr.15: 40176555-40176555 on Build GRCh38
rs77520855 Chr.15: 40176576-40176576 on Build GRCh38
rs28989186 Chr.15: 40176672-40176672 on Build GRCh38
rs145028054 Chr.15: 40176642-40176642 on Build GRCh38
rs567916146 Chr.15: 40176673-40176673 on Build GRCh38
rs758667204 Chr.15: 40176575-40176575 on Build GRCh38
rs779639976 Chr.15: 40176659-40176659 on Build GRCh38
rs375798678 Chr.15: 40176634-40176634 on Build GRCh38
rs138809057 Chr.15: 40176665-40176665 on Build GRCh38
rs587778145 Chr.15: 40176664-40176664 on Build GRCh38
rs751855402 Chr.15: 40176583-40176583 on Build GRCh38
rs139794931 Chr.15: 40176647-40176647 on Build GRCh38
rs761648006 Chr.15: 40176668-40176668 on Build GRCh38
rs889281728 Chr.15: 40176633-40176633 on Build GRCh38
rs781338363 Chr.15: 40176593-40176593 on Build GRCh38

View Full Product Details