Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233760262-233760420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT1A1
Entrez Gene ID
54658
Gene Name
UDP glucuronosyltransferase family 1 member A1
Gene Aliases
BILIQTL1, GNT1, HUG-BR1, UDPGT, UDPGT 1-1, UGT1, UGT1A
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233760273-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A10
Entrez Gene ID
54575
Gene Name
UDP glucuronosyltransferase family 1 member A10
Gene Aliases
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233636477-233773305 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A3
Entrez Gene ID
54659
Gene Name
UDP glucuronosyltransferase family 1 member A3
Gene Aliases
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233729127-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A4
Entrez Gene ID
54657
Gene Name
UDP glucuronosyltransferase family 1 member A4
Gene Aliases
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233718792-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A5
Entrez Gene ID
54579
Gene Name
UDP glucuronosyltransferase family 1 member A5
Gene Aliases
UDPGT, UDPGT 1-5, UGT1E
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233712992-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A6
Entrez Gene ID
54578
Gene Name
UDP glucuronosyltransferase family 1 member A6
Gene Aliases
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233691675-233773305 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233681938-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A8
Entrez Gene ID
54576
Gene Name
UDP glucuronosyltransferase family 1 member A8
Gene Aliases
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233617645-233773299 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A9
Entrez Gene ID
54600
Gene Name
UDP glucuronosyltransferase family 1 member A9
Gene Aliases
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233671898-233773305 on Build GRCh38
Transcripts
NM_000463.2 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4467647 c.14C>T p.S5F Chr.2: 233760301-233760301 on Build GRCh38
6026532 c.72G>A p.V24V Chr.2: 233760359-233760359 on Build GRCh38
7255341 c.39G>A p.L13L Chr.2: 233760326-233760326 on Build GRCh38
3748121 c.84G>A p.G28G Chr.2: 233760371-233760371 on Build GRCh38
7255340 c.39G>A p.L13L Chr.2: 233760326-233760326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752018052 Chr.2: 233760284-233760284 on Build GRCh38
rs1006073064 Chr.2: 233760306-233760306 on Build GRCh38
rs772038779 Chr.2: 233760317-233760317 on Build GRCh38
rs764918207 Chr.2: 233760359-233760359 on Build GRCh38
rs368348566 Chr.2: 233760385-233760385 on Build GRCh38
rs897355036 Chr.2: 233760344-233760344 on Build GRCh38
rs112008667 Chr.2: 233760283-233760283 on Build GRCh38
rs745957787 Chr.2: 233760305-233760305 on Build GRCh38
rs1032753915 Chr.2: 233760375-233760375 on Build GRCh38
rs751894919 Chr.2: 233760360-233760360 on Build GRCh38
rs201984525 Chr.2: 233760319-233760319 on Build GRCh38
rs775463336 Chr.2: 233760365-233760378 on Build GRCh38
rs74720349 Chr.2: 233760295-233760295 on Build GRCh38
rs995864584 Chr.2: 233760371-233760371 on Build GRCh38
rs780016114 Chr.2: 233760397-233760397 on Build GRCh38
rs111033541 Chr.2: 233760331-233760331 on Build GRCh38
rs759212811 Chr.2: 233760348-233760348 on Build GRCh38
rs781590934 Chr.2: 233760299-233760299 on Build GRCh38
rs780253764 Chr.2: 233760308-233760308 on Build GRCh38
rs72551339 Chr.2: 233760402-233760402 on Build GRCh38
rs386656374 Chr.2: 233760312-233760313 on Build GRCh38
rs768521253 Chr.2: 233760310-233760310 on Build GRCh38
rs994298046 Chr.2: 233760365-233760365 on Build GRCh38
rs761736540 Chr.2: 233760313-233760313 on Build GRCh38
rs375204962 Chr.2: 233760376-233760376 on Build GRCh38
rs765814343 Chr.2: 233760329-233760329 on Build GRCh38
rs768058050 Chr.2: 233760380-233760380 on Build GRCh38
rs1018382617 Chr.2: 233760398-233760398 on Build GRCh38
rs760176104 Chr.2: 233760323-233760323 on Build GRCh38
rs756175438 Chr.2: 233760387-233760387 on Build GRCh38
rs376515645 Chr.2: 233760285-233760285 on Build GRCh38
rs753493472 Chr.2: 233760338-233760338 on Build GRCh38
rs749552053 Chr.2: 233760309-233760309 on Build GRCh38
rs370790922 Chr.2: 233760312-233760312 on Build GRCh38
rs1029837369 Chr.2: 233760383-233760383 on Build GRCh38

View Full Product Details