Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33319353-33319509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAXX
Entrez Gene ID
1616
Gene Name
death domain associated protein
Gene Aliases
BING2, DAP6, EAP1
UniGene
Hs.336916
Gene Chromosome Location
Chr.6: 33318558-33323016 on Build GRCh38
Transcripts
NM_001141969.1 NM_001141970.1 NM_001254717.1 NM_001350.4 XM_005248860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421364 c.1897C>T p.R633W Chr.6: 33319423-33319423 on Build GRCh38
6971866 c.1903G>C p.E635Q Chr.6: 33319417-33319417 on Build GRCh38
1443783 c.1884_1885insC p.C629fs*29 Chr.6: 33319435-33319436 on Build GRCh38
1443784 c.1884delC p.C629fs*16 Chr.6: 33319436-33319436 on Build GRCh38
6503461 c.1852_1852delT p.S618fs*27 Chr.6: 33319468-33319468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764773519 Chr.6: 33319470-33319470 on Build GRCh38
rs751278404 Chr.6: 33319488-33319488 on Build GRCh38
rs1031681871 Chr.6: 33319447-33319447 on Build GRCh38
rs2051625 Chr.6: 33319406-33319406 on Build GRCh38
rs568745117 Chr.6: 33319440-33319440 on Build GRCh38
rs764000410 Chr.6: 33319484-33319484 on Build GRCh38
rs376777425 Chr.6: 33319421-33319421 on Build GRCh38
rs780221003 Chr.6: 33319391-33319391 on Build GRCh38
rs981409816 Chr.6: 33319422-33319422 on Build GRCh38
rs550192421 Chr.6: 33319438-33319438 on Build GRCh38
rs746991098 Chr.6: 33319385-33319385 on Build GRCh38
rs148540788 Chr.6: 33319471-33319471 on Build GRCh38
rs200527264 Chr.6: 33319411-33319411 on Build GRCh38
rs956162512 Chr.6: 33319473-33319473 on Build GRCh38
rs757117707 Chr.6: 33319386-33319386 on Build GRCh38
rs762649417 Chr.6: 33319479-33319479 on Build GRCh38
rs980817638 Chr.6: 33319419-33319419 on Build GRCh38
rs773398360 Chr.6: 33319423-33319423 on Build GRCh38
rs528956295 Chr.6: 33319413-33319413 on Build GRCh38
rs200566552 Chr.6: 33319441-33319441 on Build GRCh38
rs759286823 Chr.6: 33319397-33319398 on Build GRCh38
rs748299232 Chr.6: 33319415-33319415 on Build GRCh38

View Full Product Details