Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98890321-98890481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRRAP
Entrez Gene ID
8295
Gene Name
transformation/transcription domain associated protein
Gene Aliases
PAF350/400, PAF400, STAF40, TR-AP, Tra1
UniGene
Hs.203952
Gene Chromosome Location
Chr.7: 98878490-99013243 on Build GRCh38
Transcripts
NM_001244580.1 NM_003496.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5886553 c.199C>T p.P67S Chr.7: 98890383-98890383 on Build GRCh38
248027 c.155T>C p.V52A Chr.7: 98890339-98890339 on Build GRCh38
5768663 c.170A>G p.Q57R Chr.7: 98890354-98890354 on Build GRCh38
7128040 c.158C>A p.T53K Chr.7: 98890342-98890342 on Build GRCh38
4799310 c.151-1G>A p.? Chr.7: 98890334-98890334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905910952 Chr.7: 98890453-98890453 on Build GRCh38
rs782125130 Chr.7: 98890361-98890361 on Build GRCh38
rs782732311 Chr.7: 98890403-98890403 on Build GRCh38
rs74478141 Chr.7: 98890366-98890366 on Build GRCh38
rs782352971 Chr.7: 98890364-98890364 on Build GRCh38
rs782429150 Chr.7: 98890388-98890388 on Build GRCh38
rs782273085 Chr.7: 98890451-98890451 on Build GRCh38
rs782566165 Chr.7: 98890435-98890435 on Build GRCh38
rs781887882 Chr.7: 98890430-98890430 on Build GRCh38
rs375984896 Chr.7: 98890384-98890384 on Build GRCh38
rs1045712129 Chr.7: 98890378-98890378 on Build GRCh38
rs782633969 Chr.7: 98890439-98890439 on Build GRCh38
rs143708233 Chr.7: 98890368-98890368 on Build GRCh38
rs781841596 Chr.7: 98890370-98890370 on Build GRCh38

View Full Product Details