Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177091697-177091859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR4
Entrez Gene ID
2264
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
CD334, JTK2, TKF
UniGene
Hs.165950
Gene Chromosome Location
Chr.5: 177086872-177098144 on Build GRCh38
Transcripts
NM_001291980.1 NM_002011.4 NM_022963.3 NM_213647.2 XM_005265838.3 XM_011534464.2 XM_011534465.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5608833 c.652G>A p.D218N Chr.5: 177091733-177091733 on Build GRCh38
6955017 c.700C>T p.R234C Chr.5: 177091781-177091781 on Build GRCh38
6925981 c.633G>A p.M211I Chr.5: 177091714-177091714 on Build GRCh38
6953295 c.679G>C p.E227Q Chr.5: 177091760-177091760 on Build GRCh38
6198041 c.697A>G p.I233V Chr.5: 177091778-177091778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772846686 Chr.5: 177091819-177091819 on Build GRCh38
rs764174482 Chr.5: 177091732-177091732 on Build GRCh38
rs775100440 Chr.5: 177091721-177091721 on Build GRCh38
rs767200148 Chr.5: 177091817-177091817 on Build GRCh38
rs752817668 Chr.5: 177091781-177091781 on Build GRCh38
rs753812223 Chr.5: 177091736-177091736 on Build GRCh38
rs373098233 Chr.5: 177091837-177091837 on Build GRCh38
rs146983133 Chr.5: 177091726-177091726 on Build GRCh38
rs452885 Chr.5: 177091783-177091783 on Build GRCh38
rs760145859 Chr.5: 177091728-177091728 on Build GRCh38
rs771333444 Chr.5: 177091793-177091793 on Build GRCh38
rs764871795 Chr.5: 177091761-177091761 on Build GRCh38
rs771891900 Chr.5: 177091808-177091808 on Build GRCh38
rs3135923 Chr.5: 177091765-177091765 on Build GRCh38
rs962807397 Chr.5: 177091731-177091731 on Build GRCh38
rs374900591 Chr.5: 177091816-177091816 on Build GRCh38
rs781285001 Chr.5: 177091767-177091767 on Build GRCh38
rs200360247 Chr.5: 177091825-177091825 on Build GRCh38
rs199935139 Chr.5: 177091782-177091782 on Build GRCh38
rs200232395 Chr.5: 177091737-177091737 on Build GRCh38
rs369336618 Chr.5: 177091814-177091814 on Build GRCh38
rs924965465 Chr.5: 177091824-177091824 on Build GRCh38
rs201851606 Chr.5: 177091810-177091810 on Build GRCh38
rs755103491 Chr.5: 177091766-177091766 on Build GRCh38
rs373275390 Chr.5: 177091815-177091815 on Build GRCh38
rs749756260 Chr.5: 177091785-177091785 on Build GRCh38
rs762709285 Chr.5: 177091820-177091820 on Build GRCh38
rs975844209 Chr.5: 177091720-177091720 on Build GRCh38
rs775260233 Chr.5: 177091809-177091809 on Build GRCh38
rs747857755 Chr.5: 177091829-177091830 on Build GRCh38
rs779090527 Chr.5: 177091795-177091795 on Build GRCh38
rs376741183 Chr.5: 177091786-177091786 on Build GRCh38
rs745557685 Chr.5: 177091800-177091800 on Build GRCh38

View Full Product Details