Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 225825-225973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6940153 c.457-1G>T p.? Chr.5: 225882-225882 on Build GRCh38
4849570 c.485G>C p.R162T Chr.5: 225911-225911 on Build GRCh38
5809763 c.511C>T p.R171C Chr.5: 225937-225937 on Build GRCh38
6942838 c.490G>C p.E164Q Chr.5: 225916-225916 on Build GRCh38
3854557 c.515C>T p.A172V Chr.5: 225941-225941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762379590 Chr.5: 225862-225863 on Build GRCh38
rs774589410 Chr.5: 225893-225893 on Build GRCh38
rs371775474 Chr.5: 225867-225867 on Build GRCh38
rs777873911 Chr.5: 225887-225887 on Build GRCh38
rs749824479 Chr.5: 225890-225890 on Build GRCh38
rs181278759 Chr.5: 225942-225942 on Build GRCh38
rs765899092 Chr.5: 225875-225875 on Build GRCh38
rs763578369 Chr.5: 225925-225925 on Build GRCh38
rs750177090 Chr.5: 225861-225861 on Build GRCh38
rs765157205 Chr.5: 225939-225939 on Build GRCh38
rs753004001 Chr.5: 225949-225949 on Build GRCh38
rs766258363 Chr.5: 225946-225946 on Build GRCh38
rs989318548 Chr.5: 225894-225894 on Build GRCh38
rs770052391 Chr.5: 225879-225879 on Build GRCh38
rs189243489 Chr.5: 225855-225855 on Build GRCh38
rs569384870 Chr.5: 225892-225892 on Build GRCh38
rs375067326 Chr.5: 225914-225914 on Build GRCh38
rs587782076 Chr.5: 225938-225938 on Build GRCh38
rs759827541 Chr.5: 225902-225902 on Build GRCh38
rs776193478 Chr.5: 225937-225937 on Build GRCh38
rs775074528 Chr.5: 225904-225904 on Build GRCh38
rs778632786 Chr.5: 225860-225860 on Build GRCh38
rs755259343 Chr.5: 225866-225866 on Build GRCh38
rs878854632 Chr.5: 225881-225883 on Build GRCh38
rs748265289 Chr.5: 225872-225872 on Build GRCh38
rs771741537 Chr.5: 225903-225903 on Build GRCh38

View Full Product Details