Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 78040069-78040328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS24
Entrez Gene ID
6229
Gene Name
ribosomal protein S24
Gene Aliases
DBA3, S24
UniGene
Hs.280130
Gene Chromosome Location
Chr.10: 78033760-78056813 on Build GRCh38
Transcripts
NM_001026.4 NM_001142282.1 NM_001142283.1 NM_001142284.1 NM_001142285.1 NM_033022.3

SNP Detail

SNP ID Chromosome Location
rs369522816 Chr.10: 78040198-78040198 on Build GRCh38
rs996328102 Chr.10: 78040260-78040260 on Build GRCh38
rs776720464 Chr.10: 78040209-78040209 on Build GRCh38
rs369715779 Chr.10: 78040173-78040174 on Build GRCh38
rs956789963 Chr.10: 78040166-78040166 on Build GRCh38
rs754719579 Chr.10: 78040167-78040167 on Build GRCh38
rs751517665 Chr.10: 78040177-78040178 on Build GRCh38
rs749284970 Chr.10: 78040126-78040126 on Build GRCh38
rs1031105613 Chr.10: 78040155-78040158 on Build GRCh38
rs766642493 Chr.10: 78040243-78040243 on Build GRCh38
rs1014571168 Chr.10: 78040290-78040290 on Build GRCh38
rs749288109 Chr.10: 78040193-78040193 on Build GRCh38
rs770817159 Chr.10: 78040195-78040195 on Build GRCh38
rs190500272 Chr.10: 78040284-78040284 on Build GRCh38
rs759908455 Chr.10: 78040254-78040254 on Build GRCh38
rs747183790 Chr.10: 78040200-78040200 on Build GRCh38
rs200803982 Chr.10: 78040226-78040226 on Build GRCh38
rs115588031 Chr.10: 78040285-78040285 on Build GRCh38
rs930990648 Chr.10: 78040236-78040236 on Build GRCh38
rs755303994 Chr.10: 78040249-78040249 on Build GRCh38
rs780725121 Chr.10: 78040181-78040181 on Build GRCh38
rs765319634 Chr.10: 78040223-78040223 on Build GRCh38
rs758709816 Chr.10: 78040151-78040151 on Build GRCh38
rs201639485 Chr.10: 78040190-78040190 on Build GRCh38
rs7899453 Chr.10: 78040201-78040201 on Build GRCh38
rs766878719 Chr.10: 78040201-78040201 on Build GRCh38
rs139289215 Chr.10: 78040217-78040217 on Build GRCh38
rs761371504 Chr.10: 78040181-78040182 on Build GRCh38
rs149363889 Chr.10: 78040283-78040283 on Build GRCh38
rs774268458 Chr.10: 78040197-78040197 on Build GRCh38
rs751171895 Chr.10: 78040152-78040152 on Build GRCh38
rs747628505 Chr.10: 78040186-78040186 on Build GRCh38
rs147296007 Chr.10: 78040170-78040170 on Build GRCh38
rs564427523 Chr.10: 78040150-78040150 on Build GRCh38
rs1040629310 Chr.10: 78040214-78040214 on Build GRCh38
rs1049297530 Chr.10: 78040281-78040281 on Build GRCh38
rs527254820 Chr.10: 78040244-78040244 on Build GRCh38
rs751895913 Chr.10: 78040245-78040245 on Build GRCh38
rs763522166 Chr.10: 78040242-78040242 on Build GRCh38

View Full Product Details