Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45419343-45419616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
UniGene
Hs.437388
Gene Chromosome Location
Chr.20: 45416067-45426245 on Build GRCh38
Transcripts
NM_001184728.2 NM_001184729.2 NM_001184730.2 NM_015937.5 NR_047691.1 NR_047692.1 NR_047693.1 NR_047694.1 NR_047695.1 XM_005260430.2 XM_005260432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102215 c.603C>G p.L201L Chr.20: 45419512-45419512 on Build GRCh38
7078925 c.615G>A p.L205L Chr.20: 45419524-45419524 on Build GRCh38
1027248 c.550G>A p.E184K Chr.20: 45419351-45419351 on Build GRCh38
1326715 c.623A>G p.D208G Chr.20: 45419532-45419532 on Build GRCh38
7423072 c.631T>C p.F211L Chr.20: 45419540-45419540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751421039 Chr.20: 45419399-45419399 on Build GRCh38
rs772917338 Chr.20: 45419446-45419446 on Build GRCh38
rs200144107 Chr.20: 45419425-45419425 on Build GRCh38
rs559812381 Chr.20: 45419576-45419576 on Build GRCh38
rs778998943 Chr.20: 45419484-45419484 on Build GRCh38
rs369525139 Chr.20: 45419580-45419580 on Build GRCh38
rs755345721 Chr.20: 45419427-45419428 on Build GRCh38
rs755869697 Chr.20: 45419550-45419550 on Build GRCh38
rs763833981 Chr.20: 45419556-45419556 on Build GRCh38
rs772970201 Chr.20: 45419516-45419516 on Build GRCh38
rs757356505 Chr.20: 45419483-45419483 on Build GRCh38
rs372801153 Chr.20: 45419439-45419439 on Build GRCh38
rs147475258 Chr.20: 45419584-45419584 on Build GRCh38
rs919248328 Chr.20: 45419456-45419456 on Build GRCh38
rs746342605 Chr.20: 45419426-45419426 on Build GRCh38
rs756261965 Chr.20: 45419411-45419411 on Build GRCh38
rs749354579 Chr.20: 45419420-45419420 on Build GRCh38
rs772097684 Chr.20: 45419495-45419497 on Build GRCh38
rs769175313 Chr.20: 45419593-45419593 on Build GRCh38
rs775787608 Chr.20: 45419506-45419506 on Build GRCh38
rs1035548150 Chr.20: 45419423-45419423 on Build GRCh38
rs752751937 Chr.20: 45419367-45419367 on Build GRCh38
rs536857924 Chr.20: 45419480-45419480 on Build GRCh38
rs753460333 Chr.20: 45419364-45419364 on Build GRCh38
rs933610455 Chr.20: 45419460-45419460 on Build GRCh38
rs542261911 Chr.20: 45419423-45419424 on Build GRCh38
rs561526151 Chr.20: 45419424-45419425 on Build GRCh38
rs754895316 Chr.20: 45419451-45419451 on Build GRCh38
rs757408011 Chr.20: 45419572-45419572 on Build GRCh38
rs779325905 Chr.20: 45419428-45419439 on Build GRCh38
rs747421123 Chr.20: 45419590-45419590 on Build GRCh38
rs376282102 Chr.20: 45419454-45419454 on Build GRCh38
rs369133115 Chr.20: 45419419-45419419 on Build GRCh38
rs3092108 Chr.20: 45419425-45419426 on Build GRCh38
rs759457479 Chr.20: 45419534-45419534 on Build GRCh38
rs780128113 Chr.20: 45419396-45419396 on Build GRCh38
rs371656121 Chr.20: 45419401-45419401 on Build GRCh38
rs758888947 Chr.20: 45419577-45419577 on Build GRCh38
rs764979870 Chr.20: 45419443-45419443 on Build GRCh38
rs752531067 Chr.20: 45419463-45419463 on Build GRCh38
rs777947109 Chr.20: 45419472-45419472 on Build GRCh38
rs368012737 Chr.20: 45419447-45419447 on Build GRCh38
rs749890287 Chr.20: 45419381-45419381 on Build GRCh38
rs991584570 Chr.20: 45419431-45419431 on Build GRCh38
rs756384737 Chr.20: 45419467-45419467 on Build GRCh38
rs397828499 Chr.20: 45419431-45419432 on Build GRCh38
rs754077727 Chr.20: 45419566-45419566 on Build GRCh38
rs186550658 Chr.20: 45419403-45419403 on Build GRCh38
rs749407608 Chr.20: 45419482-45419482 on Build GRCh38
rs143235179 Chr.20: 45419535-45419535 on Build GRCh38
rs772567664 Chr.20: 45419500-45419500 on Build GRCh38
rs751482979 Chr.20: 45419449-45419449 on Build GRCh38
rs140897889 Chr.20: 45419522-45419522 on Build GRCh38
rs574183358 Chr.20: 45419543-45419543 on Build GRCh38
rs543550769 Chr.20: 45419448-45419448 on Build GRCh38
rs371987730 Chr.20: 45419425-45419425 on Build GRCh38
rs777937565 Chr.20: 45419515-45419516 on Build GRCh38
rs375333648 Chr.20: 45419404-45419404 on Build GRCh38
rs747368010 Chr.20: 45419509-45419509 on Build GRCh38
rs201614605 Chr.20: 45419490-45419490 on Build GRCh38
rs201045122 Chr.20: 45419573-45419573 on Build GRCh38
rs201798860 Chr.20: 45419424-45419424 on Build GRCh38
rs144409753 Chr.20: 45419521-45419521 on Build GRCh38
rs781001584 Chr.20: 45419402-45419402 on Build GRCh38
rs761482087 Chr.20: 45419374-45419374 on Build GRCh38
rs772341021 Chr.20: 45419432-45419432 on Build GRCh38
rs778406276 Chr.20: 45419494-45419494 on Build GRCh38
rs758480164 Chr.20: 45419382-45419382 on Build GRCh38

View Full Product Details