Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240717373-240717628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
UniGene
Hs.516802
Gene Chromosome Location
Chr.2: 240713764-240821025 on Build GRCh38
Transcripts
NM_001244008.1 NM_001320705.1 NM_004321.6 XM_005247022.1 XM_005247024.1 XM_005247027.1 XM_005247028.2 XM_006712605.1 XM_011511364.1 XM_011511365.1 XM_011511366.2 XM_011511367.1 XM_017004383.1 XM_017004384.1 XM_017004385.1 XM_017004386.1 XM_017004387.1 XM_017004388.1 XM_017004389.1 XM_017004390.1 XM_017004391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6091211 c.5354G>T p.R1785M Chr.2: 240717386-240717386 on Build GRCh38
1255799 c.5059C>A p.Q1687K Chr.2: 240717378-240717378 on Build GRCh38
6091212 c.5051G>T p.R1684M Chr.2: 240717386-240717386 on Build GRCh38
6157881 c.5043C>A p.S1681S Chr.2: 240717394-240717394 on Build GRCh38
6157880 c.5346C>A p.S1782S Chr.2: 240717394-240717394 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003986227 Chr.2: 240717522-240717522 on Build GRCh38
rs758806789 Chr.2: 240717410-240717410 on Build GRCh38
rs1027993154 Chr.2: 240717557-240717557 on Build GRCh38
rs769679424 Chr.2: 240717439-240717439 on Build GRCh38
rs747320289 Chr.2: 240717428-240717428 on Build GRCh38
rs189282597 Chr.2: 240717469-240717469 on Build GRCh38
rs1001759107 Chr.2: 240717413-240717413 on Build GRCh38
rs907521890 Chr.2: 240717603-240717603 on Build GRCh38
rs534411093 Chr.2: 240717486-240717486 on Build GRCh38
rs986874899 Chr.2: 240717477-240717477 on Build GRCh38
rs1007989919 Chr.2: 240717578-240717578 on Build GRCh38
rs898410979 Chr.2: 240717536-240717536 on Build GRCh38
rs558374091 Chr.2: 240717606-240717606 on Build GRCh38
rs777631361 Chr.2: 240717559-240717559 on Build GRCh38
rs905642704 Chr.2: 240717399-240717399 on Build GRCh38
rs1019361196 Chr.2: 240717581-240717581 on Build GRCh38
rs964200437 Chr.2: 240717491-240717491 on Build GRCh38
rs546283159 Chr.2: 240717492-240717492 on Build GRCh38
rs920102677 Chr.2: 240717513-240717513 on Build GRCh38
rs369622428 Chr.2: 240717429-240717429 on Build GRCh38
rs149656076 Chr.2: 240717604-240717604 on Build GRCh38
rs889424629 Chr.2: 240717566-240717566 on Build GRCh38
rs748416328 Chr.2: 240717401-240717401 on Build GRCh38
rs575986280 Chr.2: 240717586-240717586 on Build GRCh38
rs776337224 Chr.2: 240717467-240717467 on Build GRCh38
rs181750425 Chr.2: 240717496-240717496 on Build GRCh38
rs764018540 Chr.2: 240717456-240717456 on Build GRCh38
rs776738212 Chr.2: 240717431-240717431 on Build GRCh38
rs995796841 Chr.2: 240717540-240717540 on Build GRCh38
rs1046890706 Chr.2: 240717497-240717497 on Build GRCh38
rs775518990 Chr.2: 240717441-240717441 on Build GRCh38
rs535790499 Chr.2: 240717521-240717521 on Build GRCh38
rs777787746 Chr.2: 240717422-240717422 on Build GRCh38
rs570255531 Chr.2: 240717481-240717481 on Build GRCh38
rs780821029 Chr.2: 240717479-240717479 on Build GRCh38
rs557605169 Chr.2: 240717552-240717552 on Build GRCh38
rs371994677 Chr.2: 240717432-240717432 on Build GRCh38
rs983015531 Chr.2: 240717524-240717524 on Build GRCh38

View Full Product Details