Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 23203658-23203931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGCG
Entrez Gene ID
6445
Gene Name
sarcoglycan gamma
Gene Aliases
35DAG, A4, DAGA4, DMDA, DMDA1, LGMD2C, MAM, SCARMD2, SCG3, gamma-SG
UniGene
Hs.37167
Gene Chromosome Location
Chr.13: 23160308-23325165 on Build GRCh38
Transcripts
NM_000231.2 XM_005266505.2 XM_006719861.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1287854 c.25G>A p.A9T Chr.13: 23203719-23203719 on Build GRCh38
336024 c.73A>G p.K25E Chr.13: 23203767-23203767 on Build GRCh38
945981 c.57G>T p.E19D Chr.13: 23203751-23203751 on Build GRCh38
945980 c.31G>T p.E11* Chr.13: 23203725-23203725 on Build GRCh38
3467676 c.168G>A p.W56* Chr.13: 23203862-23203862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369851744 Chr.13: 23203688-23203688 on Build GRCh38
rs140310896 Chr.13: 23203701-23203701 on Build GRCh38
rs778502285 Chr.13: 23203793-23203793 on Build GRCh38
rs138880406 Chr.13: 23203851-23203851 on Build GRCh38
rs1022001256 Chr.13: 23203814-23203814 on Build GRCh38
rs546543109 Chr.13: 23203740-23203740 on Build GRCh38
rs775643720 Chr.13: 23203805-23203805 on Build GRCh38
rs771657671 Chr.13: 23203716-23203716 on Build GRCh38
rs762777463 Chr.13: 23203780-23203781 on Build GRCh38
rs532463691 Chr.13: 23203894-23203894 on Build GRCh38
rs746698647 Chr.13: 23203896-23203896 on Build GRCh38
rs919571338 Chr.13: 23203874-23203874 on Build GRCh38
rs797045106 Chr.13: 23203893-23203896 on Build GRCh38
rs771255124 Chr.13: 23203798-23203798 on Build GRCh38
rs754055956 Chr.13: 23203708-23203708 on Build GRCh38
rs747860532 Chr.13: 23203713-23203713 on Build GRCh38
rs373083509 Chr.13: 23203860-23203860 on Build GRCh38
rs886043151 Chr.13: 23203771-23203771 on Build GRCh38
rs780186997 Chr.13: 23203790-23203790 on Build GRCh38
rs202078221 Chr.13: 23203775-23203775 on Build GRCh38
rs1002721062 Chr.13: 23203684-23203684 on Build GRCh38
rs369936288 Chr.13: 23203836-23203836 on Build GRCh38
rs747549369 Chr.13: 23203794-23203794 on Build GRCh38
rs745599370 Chr.13: 23203687-23203687 on Build GRCh38
rs764748199 Chr.13: 23203721-23203721 on Build GRCh38
rs764468720 Chr.13: 23203812-23203812 on Build GRCh38
rs751808758 Chr.13: 23203827-23203827 on Build GRCh38
rs1031067187 Chr.13: 23203778-23203778 on Build GRCh38
rs373335254 Chr.13: 23203692-23203692 on Build GRCh38
rs376410504 Chr.13: 23203760-23203760 on Build GRCh38
rs764286219 Chr.13: 23203690-23203690 on Build GRCh38
rs777445740 Chr.13: 23203893-23203893 on Build GRCh38
rs761027747 Chr.13: 23203728-23203728 on Build GRCh38
rs200502077 Chr.13: 23203890-23203890 on Build GRCh38
rs763248287 Chr.13: 23203752-23203752 on Build GRCh38
rs76916029 Chr.13: 23203788-23203788 on Build GRCh38
rs754949325 Chr.13: 23203837-23203837 on Build GRCh38
rs768187079 Chr.13: 23203706-23203706 on Build GRCh38
rs148041867 Chr.13: 23203711-23203711 on Build GRCh38
rs35105771 Chr.13: 23203702-23203702 on Build GRCh38
rs760108586 Chr.13: 23203799-23203799 on Build GRCh38
rs770141199 Chr.13: 23203804-23203804 on Build GRCh38
rs781760379 Chr.13: 23203852-23203852 on Build GRCh38
rs752669226 Chr.13: 23203751-23203751 on Build GRCh38
rs773575178 Chr.13: 23203720-23203720 on Build GRCh38
rs886044348 Chr.13: 23203875-23203875 on Build GRCh38
rs1800349 Chr.13: 23203806-23203806 on Build GRCh38
rs752310028 Chr.13: 23203866-23203866 on Build GRCh38
rs546376689 Chr.13: 23203869-23203869 on Build GRCh38
rs999585272 Chr.13: 23203776-23203776 on Build GRCh38
rs755404457 Chr.13: 23203795-23203795 on Build GRCh38
rs1019389043 Chr.13: 23203831-23203831 on Build GRCh38
rs886042587 Chr.13: 23203884-23203884 on Build GRCh38
rs1020872078 Chr.13: 23203741-23203741 on Build GRCh38
rs140810408 Chr.13: 23203796-23203796 on Build GRCh38
rs753775490 Chr.13: 23203792-23203792 on Build GRCh38
rs765392154 Chr.13: 23203750-23203750 on Build GRCh38
rs886043940 Chr.13: 23203855-23203855 on Build GRCh38
rs759576122 Chr.13: 23203746-23203746 on Build GRCh38

View Full Product Details