Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 101063159-101063430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3CA
Entrez Gene ID
5530
Gene Name
protein phosphatase 3 catalytic subunit alpha
Gene Aliases
CALN, CALNA, CALNA1, CCN1, CNA1, PPP2B
UniGene
Hs.435512
Gene Chromosome Location
Chr.4: 101023430-101347471 on Build GRCh38
Transcripts
NM_000944.4 NM_001130691.1 NM_001130692.1 XM_017008365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016063 c.964_965insAT p.L322fs*2 Chr.4: 101063348-101063349 on Build GRCh38
4612167 c.1013_1015delCTC p.P338delP Chr.4: 101063298-101063300 on Build GRCh38
4625206 c.1025G>T p.W342L Chr.4: 101063288-101063288 on Build GRCh38
5918489 c.1036T>A p.F346I Chr.4: 101063277-101063277 on Build GRCh38
3598679 c.1018C>T p.P340S Chr.4: 101063295-101063295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201499740 Chr.4: 101063202-101063202 on Build GRCh38
rs771970265 Chr.4: 101063212-101063212 on Build GRCh38
rs779008232 Chr.4: 101063386-101063386 on Build GRCh38
rs748137607 Chr.4: 101063211-101063211 on Build GRCh38
rs200757163 Chr.4: 101063275-101063275 on Build GRCh38
rs768159553 Chr.4: 101063352-101063352 on Build GRCh38
rs940015493 Chr.4: 101063191-101063191 on Build GRCh38
rs750904321 Chr.4: 101063374-101063374 on Build GRCh38
rs755209181 Chr.4: 101063384-101063384 on Build GRCh38
rs749531202 Chr.4: 101063382-101063382 on Build GRCh38
rs748260655 Chr.4: 101063396-101063396 on Build GRCh38
rs756441434 Chr.4: 101063377-101063377 on Build GRCh38
rs560796079 Chr.4: 101063379-101063379 on Build GRCh38
rs919155461 Chr.4: 101063228-101063228 on Build GRCh38
rs768686035 Chr.4: 101063206-101063206 on Build GRCh38
rs760412015 Chr.4: 101063261-101063261 on Build GRCh38
rs997375921 Chr.4: 101063198-101063198 on Build GRCh38
rs776210956 Chr.4: 101063271-101063271 on Build GRCh38
rs752223096 Chr.4: 101063309-101063309 on Build GRCh38
rs780155809 Chr.4: 101063197-101063197 on Build GRCh38
rs764698335 Chr.4: 101063293-101063293 on Build GRCh38
rs774400784 Chr.4: 101063208-101063208 on Build GRCh38
rs746264506 Chr.4: 101063186-101063186 on Build GRCh38
rs766129389 Chr.4: 101063263-101063263 on Build GRCh38
rs1028112332 Chr.4: 101063200-101063200 on Build GRCh38
rs756449398 Chr.4: 101063194-101063194 on Build GRCh38
rs773096617 Chr.4: 101063260-101063260 on Build GRCh38
rs757940346 Chr.4: 101063332-101063332 on Build GRCh38

View Full Product Details