Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33707589-33707832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXMP4
Entrez Gene ID
11264
Gene Name
peroxisomal membrane protein 4
Gene Aliases
PMP24
UniGene
Hs.654857
Gene Chromosome Location
Chr.20: 33702744-33720330 on Build GRCh38
Transcripts
NM_007238.4 NM_183397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443677 c.527C>T p.S176F Chr.20: 33707818-33707818 on Build GRCh38
7075855 c.567C>G p.L189L Chr.20: 33707778-33707778 on Build GRCh38
7161018 c.578G>A p.S193N Chr.20: 33707767-33707767 on Build GRCh38
6092802 c.574G>T p.D192Y Chr.20: 33707771-33707771 on Build GRCh38
6021935 c.542C>T p.S181L Chr.20: 33707803-33707803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145235084 Chr.20: 33707762-33707762 on Build GRCh38
rs943159264 Chr.20: 33707628-33707628 on Build GRCh38
rs200916380 Chr.20: 33707719-33707719 on Build GRCh38
rs779024301 Chr.20: 33707720-33707720 on Build GRCh38
rs750627574 Chr.20: 33707793-33707793 on Build GRCh38
rs780052694 Chr.20: 33707688-33707688 on Build GRCh38
rs753817815 Chr.20: 33707812-33707812 on Build GRCh38
rs766624753 Chr.20: 33707811-33707811 on Build GRCh38
rs772966200 Chr.20: 33707753-33707753 on Build GRCh38
rs752754562 Chr.20: 33707662-33707662 on Build GRCh38
rs772041080 Chr.20: 33707742-33707742 on Build GRCh38
rs910397 Chr.20: 33707735-33707735 on Build GRCh38
rs762146417 Chr.20: 33707782-33707782 on Build GRCh38
rs1000293581 Chr.20: 33707807-33707807 on Build GRCh38
rs774864320 Chr.20: 33707764-33707764 on Build GRCh38
rs758120687 Chr.20: 33707805-33707805 on Build GRCh38
rs772378372 Chr.20: 33707622-33707622 on Build GRCh38
rs746707723 Chr.20: 33707754-33707754 on Build GRCh38
rs764549910 Chr.20: 33707763-33707763 on Build GRCh38
rs756230752 Chr.20: 33707802-33707802 on Build GRCh38
rs750696158 Chr.20: 33707661-33707661 on Build GRCh38
rs767911987 Chr.20: 33707790-33707790 on Build GRCh38
rs756212003 Chr.20: 33707667-33707667 on Build GRCh38
rs114550658 Chr.20: 33707700-33707700 on Build GRCh38

View Full Product Details