Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76729620-76729877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL23
Entrez Gene ID
124512
Gene Name
methyltransferase like 23
Gene Aliases
C17orf95, MRT44
UniGene
Hs.74655
Gene Chromosome Location
Chr.17: 76725874-76733881 on Build GRCh38
Transcripts
NM_001080510.4 NM_001206983.2 NM_001206984.2 NM_001206985.2 NM_001206986.2 NM_001206987.2 NM_001302703.1 NM_001302704.1 NM_001302705.1 NR_038193.2 XM_006721674.3 XM_006721675.1 XM_006721676.3 XM_006721678.3 XM_006721679.3 XM_006721680.2 XM_011524282.1 XM_017024145.1 XM_017024146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
270648 c.52_54delAGA p.R18delR Chr.17: 76729762-76729764 on Build GRCh38
1196959 c.20C>T p.A7V Chr.17: 76729730-76729730 on Build GRCh38
5475969 c.4T>C p.Y2H Chr.17: 76729714-76729714 on Build GRCh38
3932741 c.36G>A p.Q12Q Chr.17: 76729746-76729746 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537419468 Chr.17: 76729843-76729843 on Build GRCh38
rs202205410 Chr.17: 76729714-76729714 on Build GRCh38
rs777399691 Chr.17: 76729830-76729830 on Build GRCh38
rs201999820 Chr.17: 76729730-76729730 on Build GRCh38
rs896974820 Chr.17: 76729749-76729749 on Build GRCh38
rs762044347 Chr.17: 76729715-76729715 on Build GRCh38
rs993992164 Chr.17: 76729754-76729754 on Build GRCh38
rs775964216 Chr.17: 76729663-76729663 on Build GRCh38
rs761100307 Chr.17: 76729682-76729682 on Build GRCh38
rs756091392 Chr.17: 76729843-76729847 on Build GRCh38
rs372500815 Chr.17: 76729822-76729822 on Build GRCh38
rs1026852210 Chr.17: 76729771-76729771 on Build GRCh38
rs919332425 Chr.17: 76729804-76729804 on Build GRCh38
rs780707556 Chr.17: 76729794-76729794 on Build GRCh38
rs751481066 Chr.17: 76729765-76729765 on Build GRCh38
rs766331837 Chr.17: 76729764-76729764 on Build GRCh38
rs4788943 Chr.17: 76729840-76729840 on Build GRCh38
rs752127896 Chr.17: 76729823-76729823 on Build GRCh38
rs369238706 Chr.17: 76729706-76729706 on Build GRCh38
rs772292507 Chr.17: 76729828-76729828 on Build GRCh38
rs1001859214 Chr.17: 76729800-76729800 on Build GRCh38
rs368889510 Chr.17: 76729775-76729775 on Build GRCh38
rs750712124 Chr.17: 76729727-76729727 on Build GRCh38
rs768714343 Chr.17: 76729698-76729698 on Build GRCh38
rs765352928 Chr.17: 76729726-76729726 on Build GRCh38
rs1008956819 Chr.17: 76729719-76729719 on Build GRCh38
rs926219835 Chr.17: 76729679-76729679 on Build GRCh38
rs568386097 Chr.17: 76729827-76729827 on Build GRCh38
rs973676287 Chr.17: 76729766-76729766 on Build GRCh38

View Full Product Details