Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 308840-309091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
UniGene
Hs.101891
Gene Chromosome Location
Chr.19: 305573-344793 on Build GRCh38
Transcripts
NM_017550.1 XM_005259582.4 XM_006722769.3 XM_011528072.2 XM_011528073.1 XM_011528074.2 XM_011528075.2 XM_011528077.2 XM_011528081.2 XM_011528082.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172610 c.1040C>T p.S347L Chr.19: 308870-308870 on Build GRCh38
386904 c.1020C>G p.Y340* Chr.19: 308890-308890 on Build GRCh38
262021 c.1051G>A p.D351N Chr.19: 308859-308859 on Build GRCh38
3532031 c.1059C>T p.F353F Chr.19: 308851-308851 on Build GRCh38
1392270 c.1027C>A p.L343M Chr.19: 308883-308883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751550906 Chr.19: 308860-308860 on Build GRCh38
rs757065357 Chr.19: 308864-308864 on Build GRCh38
rs539460026 Chr.19: 308925-308925 on Build GRCh38
rs866631652 Chr.19: 308903-308903 on Build GRCh38
rs776595574 Chr.19: 308960-308960 on Build GRCh38
rs775609401 Chr.19: 308922-308922 on Build GRCh38
rs748781070 Chr.19: 308899-308899 on Build GRCh38
rs771494260 Chr.19: 308909-308909 on Build GRCh38
rs200873025 Chr.19: 308930-308930 on Build GRCh38
rs144035684 Chr.19: 308869-308869 on Build GRCh38
rs557699580 Chr.19: 308987-308987 on Build GRCh38
rs1019336275 Chr.19: 309011-309011 on Build GRCh38
rs757251634 Chr.19: 308932-308932 on Build GRCh38
rs755882371 Chr.19: 308937-308937 on Build GRCh38
rs777817036 Chr.19: 308859-308859 on Build GRCh38
rs199940044 Chr.19: 308918-308918 on Build GRCh38
rs913818412 Chr.19: 309062-309062 on Build GRCh38
rs763025488 Chr.19: 308929-308929 on Build GRCh38
rs139315873 Chr.19: 309012-309012 on Build GRCh38
rs763977821 Chr.19: 308959-308959 on Build GRCh38
rs796884926 Chr.19: 309056-309056 on Build GRCh38
rs546785742 Chr.19: 308998-308998 on Build GRCh38
rs1009492193 Chr.19: 308999-308999 on Build GRCh38
rs759868385 Chr.19: 308915-308915 on Build GRCh38
rs878971892 Chr.19: 308892-308892 on Build GRCh38
rs770212952 Chr.19: 309029-309029 on Build GRCh38
rs776949668 Chr.19: 308914-308914 on Build GRCh38
rs993569471 Chr.19: 309004-309004 on Build GRCh38
rs773640885 Chr.19: 308905-308905 on Build GRCh38
rs905480297 Chr.19: 308962-308962 on Build GRCh38
rs553586279 Chr.19: 309068-309068 on Build GRCh38
rs765452913 Chr.19: 308916-308916 on Build GRCh38
rs371603611 Chr.19: 308893-308893 on Build GRCh38
rs747528443 Chr.19: 308906-308906 on Build GRCh38
rs768081074 Chr.19: 308904-308904 on Build GRCh38
rs566019073 Chr.19: 308896-308896 on Build GRCh38
rs547599838 Chr.19: 308865-308865 on Build GRCh38
rs77788712 Chr.19: 308935-308935 on Build GRCh38
rs34768147 Chr.19: 309072-309073 on Build GRCh38
rs150821045 Chr.19: 308960-308960 on Build GRCh38
rs535502870 Chr.19: 309055-309055 on Build GRCh38
rs924982384 Chr.19: 309042-309042 on Build GRCh38
rs993783528 Chr.19: 308974-308974 on Build GRCh38
rs574699743 Chr.19: 309026-309026 on Build GRCh38
rs767599287 Chr.19: 308933-308933 on Build GRCh38

View Full Product Details