Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47437888-47438155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPSN
Entrez Gene ID
5913
Gene Name
receptor associated protein of the synapse
Gene Aliases
CMS11, CMS4C, FADS, RAPSYN, RNF205
UniGene
Hs.81218
Gene Chromosome Location
Chr.11: 47437757-47449178 on Build GRCh38
Transcripts
NM_005055.4 NM_032645.4 XM_005253042.3 XM_005253043.3 XM_011520252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3666339 c.1234G>T p.V412L Chr.11: 47437980-47437980 on Build GRCh38
4522983 c.1190G>A p.R397Q Chr.11: 47438024-47438024 on Build GRCh38
6395831 c.1211G>T p.R404L Chr.11: 47438003-47438003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368937475 Chr.11: 47438098-47438098 on Build GRCh38
rs565681668 Chr.11: 47438135-47438135 on Build GRCh38
rs973418711 Chr.11: 47438099-47438099 on Build GRCh38
rs937406014 Chr.11: 47438012-47438012 on Build GRCh38
rs747442833 Chr.11: 47438027-47438027 on Build GRCh38
rs575628861 Chr.11: 47438132-47438132 on Build GRCh38
rs908347742 Chr.11: 47438134-47438134 on Build GRCh38
rs757215612 Chr.11: 47438011-47438011 on Build GRCh38
rs759167710 Chr.11: 47438106-47438106 on Build GRCh38
rs954785577 Chr.11: 47438067-47438067 on Build GRCh38
rs946051835 Chr.11: 47437959-47437959 on Build GRCh38
rs528442857 Chr.11: 47438003-47438003 on Build GRCh38
rs1046873284 Chr.11: 47437912-47437912 on Build GRCh38
rs1013183854 Chr.11: 47438020-47438020 on Build GRCh38
rs977410401 Chr.11: 47437940-47437940 on Build GRCh38
rs768882267 Chr.11: 47438031-47438031 on Build GRCh38
rs867439892 Chr.11: 47438043-47438043 on Build GRCh38
rs762532220 Chr.11: 47438034-47438034 on Build GRCh38
rs879658032 Chr.11: 47437960-47437960 on Build GRCh38
rs201367093 Chr.11: 47438083-47438083 on Build GRCh38
rs985182143 Chr.11: 47438129-47438129 on Build GRCh38
rs146285971 Chr.11: 47437996-47437996 on Build GRCh38
rs931229290 Chr.11: 47438114-47438114 on Build GRCh38
rs45617144 Chr.11: 47437918-47437918 on Build GRCh38
rs530896580 Chr.11: 47438032-47438032 on Build GRCh38
rs759218351 Chr.11: 47437915-47437915 on Build GRCh38
rs774049017 Chr.11: 47438068-47438068 on Build GRCh38
rs757384978 Chr.11: 47437945-47437945 on Build GRCh38
rs528035740 Chr.11: 47437909-47437909 on Build GRCh38
rs770347768 Chr.11: 47438064-47438064 on Build GRCh38
rs77410879 Chr.11: 47438005-47438005 on Build GRCh38
rs201796294 Chr.11: 47438025-47438025 on Build GRCh38
rs886048386 Chr.11: 47437959-47437959 on Build GRCh38
rs370123138 Chr.11: 47438033-47438033 on Build GRCh38
rs376725663 Chr.11: 47438089-47438089 on Build GRCh38
rs139398367 Chr.11: 47438024-47438024 on Build GRCh38
rs772794085 Chr.11: 47438078-47438078 on Build GRCh38
rs1055000377 Chr.11: 47438021-47438021 on Build GRCh38
rs375413349 Chr.11: 47437939-47437939 on Build GRCh38
rs750941408 Chr.11: 47438004-47438004 on Build GRCh38
rs550030509 Chr.11: 47438082-47438082 on Build GRCh38
rs985630494 Chr.11: 47437917-47437917 on Build GRCh38

View Full Product Details