Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 3829817-3830062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP11
Entrez Gene ID
57097
Gene Name
poly(ADP-ribose) polymerase family member 11
Gene Aliases
ARTD11, C12orf6, MIB006
UniGene
Hs.657268
Gene Chromosome Location
Chr.12: 3808861-3873448 on Build GRCh38
Transcripts
NM_001286521.1 NM_001286522.1 NM_020367.5 NR_104461.1 XM_005253711.4 XM_011520970.1 XM_011520971.2 XM_011520973.2 XM_017019670.1 XM_017019671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200182 c.39A>G p.T13T Chr.12: 3829977-3829977 on Build GRCh38
468303 c.22T>A p.L8I Chr.12: 3829994-3829994 on Build GRCh38
6989346 c.55G>A p.D19N Chr.12: 3829961-3829961 on Build GRCh38
5462213 c.66G>A p.T22T Chr.12: 3829950-3829950 on Build GRCh38
938917 c.19G>T p.E7* Chr.12: 3829997-3829997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747414834 Chr.12: 3829934-3829934 on Build GRCh38
rs370699241 Chr.12: 3829846-3829846 on Build GRCh38
rs35288911 Chr.12: 3830019-3830020 on Build GRCh38
rs149437678 Chr.12: 3829890-3829890 on Build GRCh38
rs375960264 Chr.12: 3829889-3829889 on Build GRCh38
rs377635540 Chr.12: 3829853-3829853 on Build GRCh38
rs752281232 Chr.12: 3829885-3829885 on Build GRCh38
rs1001268814 Chr.12: 3829991-3829991 on Build GRCh38
rs752123367 Chr.12: 3829984-3829984 on Build GRCh38
rs769208780 Chr.12: 3829941-3829941 on Build GRCh38
rs762776964 Chr.12: 3829849-3829849 on Build GRCh38
rs773491295 Chr.12: 3829953-3829953 on Build GRCh38
rs148563477 Chr.12: 3829996-3829996 on Build GRCh38
rs751363235 Chr.12: 3829963-3829963 on Build GRCh38
rs199671627 Chr.12: 3830027-3830027 on Build GRCh38
rs758257341 Chr.12: 3829897-3829898 on Build GRCh38
rs75962260 Chr.12: 3829998-3829998 on Build GRCh38
rs780229696 Chr.12: 3830028-3830028 on Build GRCh38
rs1035961475 Chr.12: 3829909-3829909 on Build GRCh38
rs747424563 Chr.12: 3830007-3830007 on Build GRCh38
rs780504414 Chr.12: 3829882-3829882 on Build GRCh38
rs769023310 Chr.12: 3830015-3830015 on Build GRCh38
rs780692222 Chr.12: 3829906-3829906 on Build GRCh38
rs767509790 Chr.12: 3829977-3829977 on Build GRCh38
rs762727764 Chr.12: 3829956-3829956 on Build GRCh38
rs200615337 Chr.12: 3830003-3830003 on Build GRCh38
rs772632316 Chr.12: 3829891-3829891 on Build GRCh38
rs766107782 Chr.12: 3829958-3829958 on Build GRCh38
rs1045176424 Chr.12: 3829910-3829910 on Build GRCh38
rs202111420 Chr.12: 3829854-3829854 on Build GRCh38
rs144824076 Chr.12: 3829950-3829950 on Build GRCh38
rs896742612 Chr.12: 3829856-3829856 on Build GRCh38
rs759354996 Chr.12: 3829973-3829973 on Build GRCh38
rs781743199 Chr.12: 3830018-3830018 on Build GRCh38
rs769972983 Chr.12: 3829951-3829951 on Build GRCh38
rs755685808 Chr.12: 3829995-3829995 on Build GRCh38
rs756510557 Chr.12: 3829978-3829978 on Build GRCh38
rs756982225 Chr.12: 3830001-3830001 on Build GRCh38
rs34515229 Chr.12: 3829915-3829916 on Build GRCh38
rs142910026 Chr.12: 3829997-3829997 on Build GRCh38

View Full Product Details