Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 56094093-56094356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX7
Entrez Gene ID
64864
Gene Name
regulatory factor X7
Gene Aliases
RFXDC2
UniGene
Hs.745089
Gene Chromosome Location
Chr.15: 56090533-56247654 on Build GRCh38
Transcripts
NM_022841.5 XM_005254603.3 XM_011521925.2 XM_017022506.1 XM_017022507.1 XM_017022508.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502592 c.3551C>T p.P1184L Chr.15: 56094177-56094177 on Build GRCh38
6765495 c.3104C>T p.A1035V Chr.15: 56094333-56094333 on Build GRCh38
7139045 c.3631C>A p.H1211N Chr.15: 56094097-56094097 on Build GRCh38
3502590 c.3605C>T p.P1202L Chr.15: 56094123-56094123 on Build GRCh38
6021340 c.3197G>A p.R1066K Chr.15: 56094240-56094240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778210565 Chr.15: 56094130-56094130 on Build GRCh38
rs977271911 Chr.15: 56094183-56094183 on Build GRCh38
rs754509572 Chr.15: 56094235-56094235 on Build GRCh38
rs749534670 Chr.15: 56094133-56094133 on Build GRCh38
rs756395339 Chr.15: 56094124-56094124 on Build GRCh38
rs373798837 Chr.15: 56094159-56094159 on Build GRCh38
rs191773270 Chr.15: 56094234-56094234 on Build GRCh38
rs546916627 Chr.15: 56094181-56094181 on Build GRCh38
rs868531824 Chr.15: 56094292-56094292 on Build GRCh38
rs757534799 Chr.15: 56094226-56094226 on Build GRCh38
rs753963627 Chr.15: 56094217-56094217 on Build GRCh38
rs780470832 Chr.15: 56094258-56094258 on Build GRCh38
rs751397678 Chr.15: 56094204-56094204 on Build GRCh38
rs754757785 Chr.15: 56094210-56094210 on Build GRCh38
rs772613314 Chr.15: 56094146-56094146 on Build GRCh38
rs747438438 Chr.15: 56094158-56094158 on Build GRCh38
rs779630476 Chr.15: 56094170-56094170 on Build GRCh38
rs747528557 Chr.15: 56094295-56094295 on Build GRCh38
rs757230038 Chr.15: 56094136-56094136 on Build GRCh38
rs367881364 Chr.15: 56094211-56094211 on Build GRCh38
rs779076994 Chr.15: 56094143-56094143 on Build GRCh38
rs766486055 Chr.15: 56094186-56094186 on Build GRCh38
rs776089803 Chr.15: 56094163-56094163 on Build GRCh38
rs764613013 Chr.15: 56094166-56094166 on Build GRCh38
rs775815253 Chr.15: 56094151-56094151 on Build GRCh38
rs568839360 Chr.15: 56094336-56094336 on Build GRCh38
rs117724392 Chr.15: 56094316-56094316 on Build GRCh38
rs188534635 Chr.15: 56094206-56094206 on Build GRCh38
rs746325351 Chr.15: 56094145-56094145 on Build GRCh38
rs760500312 Chr.15: 56094121-56094129 on Build GRCh38
rs781644501 Chr.15: 56094331-56094331 on Build GRCh38

View Full Product Details