Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 81906894-81907133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL2
Entrez Gene ID
23266
Gene Name
adhesion G protein-coupled receptor L2
Gene Aliases
CIRL2, CL2, LEC1, LPHH1, LPHN2
UniGene
Hs.24212
Gene Chromosome Location
Chr.1: 81306155-81992738 on Build GRCh38
Transcripts
NM_001297704.1 NM_001297705.1 NM_001297706.1 NM_012302.3 XM_005270666.4 XM_005270668.2 XM_006710485.3 XM_006710488.2 XM_006710489.3 XM_017000782.1 XM_017000783.1 XM_017000784.1 XM_017000785.1 XM_017000786.1 XM_017000787.1 XM_017000788.1 XM_017000789.1 XM_017000790.1 XM_017000791.1 XM_017000792.1 XM_017000793.1 XM_017000794.1 XM_017000795.1 XM_017000796.1 XM_017000797.1 XM_017000798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6030421 c.158G>A p.G53D Chr.1: 81907101-81907101 on Build GRCh38
5449106 c.182G>A p.S61N Chr.1: 81907125-81907125 on Build GRCh38
7323537 c.148C>T p.R50* Chr.1: 81907091-81907091 on Build GRCh38
3492660 c.122C>A p.S41Y Chr.1: 81907065-81907065 on Build GRCh38
4990597 c.174G>A p.M58I Chr.1: 81907117-81907117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752432389 Chr.1: 81906979-81906979 on Build GRCh38
rs748207651 Chr.1: 81907088-81907088 on Build GRCh38
rs761900766 Chr.1: 81907054-81907054 on Build GRCh38
rs756787951 Chr.1: 81906993-81906993 on Build GRCh38
rs1053212348 Chr.1: 81906940-81906940 on Build GRCh38
rs188800668 Chr.1: 81906935-81906935 on Build GRCh38
rs373315157 Chr.1: 81906987-81906987 on Build GRCh38
rs775979077 Chr.1: 81907009-81907009 on Build GRCh38
rs774392019 Chr.1: 81907050-81907050 on Build GRCh38
rs893055734 Chr.1: 81907103-81907103 on Build GRCh38
rs1002799292 Chr.1: 81907002-81907002 on Build GRCh38
rs764948614 Chr.1: 81906971-81906971 on Build GRCh38
rs758131530 Chr.1: 81906984-81906984 on Build GRCh38
rs370067354 Chr.1: 81906968-81906968 on Build GRCh38
rs781770647 Chr.1: 81906994-81906994 on Build GRCh38
rs759539614 Chr.1: 81907090-81907090 on Build GRCh38
rs754777441 Chr.1: 81906969-81906969 on Build GRCh38
rs934693899 Chr.1: 81906936-81906936 on Build GRCh38
rs1040937174 Chr.1: 81906926-81906926 on Build GRCh38
rs570953325 Chr.1: 81906922-81906922 on Build GRCh38
rs375742550 Chr.1: 81906943-81906943 on Build GRCh38
rs746319816 Chr.1: 81906998-81906998 on Build GRCh38
rs200327379 Chr.1: 81907013-81907013 on Build GRCh38
rs746569620 Chr.1: 81906992-81906992 on Build GRCh38
rs1035446903 Chr.1: 81907069-81907069 on Build GRCh38
rs768961675 Chr.1: 81907017-81907017 on Build GRCh38
rs550961574 Chr.1: 81906921-81906921 on Build GRCh38
rs769956127 Chr.1: 81907099-81907099 on Build GRCh38
rs1011973565 Chr.1: 81906978-81906978 on Build GRCh38
rs749777923 Chr.1: 81907001-81907001 on Build GRCh38
rs765076851 Chr.1: 81907096-81907096 on Build GRCh38
rs767443295 Chr.1: 81906974-81906977 on Build GRCh38
rs906990631 Chr.1: 81906957-81906957 on Build GRCh38
rs770418448 Chr.1: 81907008-81907008 on Build GRCh38
rs550434713 Chr.1: 81907066-81907066 on Build GRCh38
rs762626450 Chr.1: 81907104-81907104 on Build GRCh38

View Full Product Details