Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11166425-11166698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK2
Entrez Gene ID
25959
Gene Name
KN motif and ankyrin repeat domains 2
Gene Aliases
ANKRD25, MXRA3, PPKWH, SIP
UniGene
Hs.284208
Gene Chromosome Location
Chr.19: 11164267-11197567 on Build GRCh38
Transcripts
NM_001136191.2 NM_015493.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362643 c.2549A>G p.E850G Chr.19: 11166589-11166589 on Build GRCh38
4693986 c.2567C>T p.S856L Chr.19: 11166571-11166571 on Build GRCh38
4814811 c.2542G>A p.D848N Chr.19: 11166596-11166596 on Build GRCh38
5388597 c.2548G>A p.E850K Chr.19: 11166566-11166566 on Build GRCh38
3362644 c.2525A>G p.E842G Chr.19: 11166589-11166589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770686291 Chr.19: 11166634-11166634 on Build GRCh38
rs753245161 Chr.19: 11166646-11166646 on Build GRCh38
rs766743147 Chr.19: 11166517-11166517 on Build GRCh38
rs199966367 Chr.19: 11166557-11166557 on Build GRCh38
rs527527895 Chr.19: 11166518-11166518 on Build GRCh38
rs912512407 Chr.19: 11166620-11166620 on Build GRCh38
rs761494510 Chr.19: 11166608-11166608 on Build GRCh38
rs753822110 Chr.19: 11166662-11166662 on Build GRCh38
rs947929094 Chr.19: 11166542-11166542 on Build GRCh38
rs867770253 Chr.19: 11166555-11166555 on Build GRCh38
rs574713155 Chr.19: 11166446-11166446 on Build GRCh38
rs768208743 Chr.19: 11166645-11166645 on Build GRCh38
rs1044446052 Chr.19: 11166663-11166663 on Build GRCh38
rs541161277 Chr.19: 11166487-11166487 on Build GRCh38
rs760464833 Chr.19: 11166619-11166620 on Build GRCh38
rs776194951 Chr.19: 11166605-11166605 on Build GRCh38
rs375647406 Chr.19: 11166657-11166657 on Build GRCh38
rs996981514 Chr.19: 11166668-11166668 on Build GRCh38
rs979171862 Chr.19: 11166566-11166566 on Build GRCh38
rs1048604877 Chr.19: 11166672-11166672 on Build GRCh38
rs546422721 Chr.19: 11166543-11166543 on Build GRCh38
rs962250513 Chr.19: 11166511-11166511 on Build GRCh38
rs372511483 Chr.19: 11166560-11166560 on Build GRCh38
rs548896322 Chr.19: 11166529-11166529 on Build GRCh38
rs749508261 Chr.19: 11166545-11166545 on Build GRCh38
rs984425386 Chr.19: 11166473-11166473 on Build GRCh38
rs117057052 Chr.19: 11166602-11166602 on Build GRCh38
rs756218023 Chr.19: 11166540-11166540 on Build GRCh38
rs754885746 Chr.19: 11166481-11166481 on Build GRCh38
rs774873998 Chr.19: 11166612-11166612 on Build GRCh38
rs755569233 Chr.19: 11166519-11166519 on Build GRCh38
rs763285208 Chr.19: 11166508-11166508 on Build GRCh38
rs199813944 Chr.19: 11166525-11166525 on Build GRCh38
rs1003285137 Chr.19: 11166524-11166524 on Build GRCh38
rs550073614 Chr.19: 11166569-11166569 on Build GRCh38
rs772013085 Chr.19: 11166570-11166570 on Build GRCh38
rs377116202 Chr.19: 11166609-11166609 on Build GRCh38
rs184022857 Chr.19: 11166504-11166504 on Build GRCh38
rs372922788 Chr.19: 11166528-11166528 on Build GRCh38
rs768197221 Chr.19: 11166603-11166603 on Build GRCh38
rs777918624 Chr.19: 11166544-11166544 on Build GRCh38
rs372628103 Chr.19: 11166623-11166623 on Build GRCh38
rs1052841408 Chr.19: 11166558-11166558 on Build GRCh38
rs146639208 Chr.19: 11166571-11166571 on Build GRCh38
rs4804573 Chr.19: 11166556-11166556 on Build GRCh38
rs764205846 Chr.19: 11166658-11166658 on Build GRCh38

View Full Product Details