Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35044768-35045025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
UniGene
Hs.491172
Gene Chromosome Location
Chr.13: 34942287-35672737 on Build GRCh38
Transcripts
NM_015678.4 XM_005266346.4 XM_005266347.4 XM_005266348.4 XM_006719803.3 XM_006719805.3 XM_006719806.3 XM_011535047.2 XM_017020544.1 XM_017020545.1 XM_017020546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4498864 c.529C>T p.L177F Chr.13: 35044949-35044949 on Build GRCh38
4752695 c.605T>G p.L202R Chr.13: 35045025-35045025 on Build GRCh38
4752693 c.530T>G p.L177R Chr.13: 35044950-35044950 on Build GRCh38
1629062 c.574G>A p.V192I Chr.13: 35044994-35044994 on Build GRCh38
4553054 c.579G>A p.K193K Chr.13: 35044999-35044999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555746647 Chr.13: 35044827-35044828 on Build GRCh38
rs541862840 Chr.13: 35044825-35044825 on Build GRCh38
rs201041916 Chr.13: 35044833-35044833 on Build GRCh38
rs915085653 Chr.13: 35044849-35044849 on Build GRCh38
rs887344142 Chr.13: 35044951-35044951 on Build GRCh38
rs758847547 Chr.13: 35044825-35044826 on Build GRCh38
rs546510305 Chr.13: 35044837-35044837 on Build GRCh38
rs749245471 Chr.13: 35044908-35044908 on Build GRCh38
rs200722476 Chr.13: 35044827-35044827 on Build GRCh38
rs112609962 Chr.13: 35044826-35044831 on Build GRCh38
rs544844440 Chr.13: 35044816-35044821 on Build GRCh38
rs899920354 Chr.13: 35044884-35044884 on Build GRCh38
rs769956666 Chr.13: 35044817-35044818 on Build GRCh38
rs927871201 Chr.13: 35044826-35044826 on Build GRCh38
rs750634766 Chr.13: 35044829-35044830 on Build GRCh38
rs7994430 Chr.13: 35044831-35044831 on Build GRCh38
rs758130352 Chr.13: 35044803-35044803 on Build GRCh38
rs771617158 Chr.13: 35044931-35044932 on Build GRCh38
rs935261288 Chr.13: 35044828-35044831 on Build GRCh38
rs755882838 Chr.13: 35044904-35044904 on Build GRCh38
rs7996448 Chr.13: 35044854-35044854 on Build GRCh38
rs576754023 Chr.13: 35044890-35044890 on Build GRCh38
rs796635653 Chr.13: 35044830-35044831 on Build GRCh38
rs71196573 Chr.13: 35044815-35044816 on Build GRCh38
rs72190399 Chr.13: 35044816-35044817 on Build GRCh38
rs1039850557 Chr.13: 35044855-35044855 on Build GRCh38
rs74051221 Chr.13: 35044829-35044829 on Build GRCh38
rs185606417 Chr.13: 35044962-35044962 on Build GRCh38
rs987971620 Chr.13: 35044839-35044839 on Build GRCh38
rs544018430 Chr.13: 35044905-35044905 on Build GRCh38
rs182264120 Chr.13: 35044927-35044927 on Build GRCh38
rs372735958 Chr.13: 35044830-35044835 on Build GRCh38

View Full Product Details