Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86466538-86466807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7176414 c.1424C>T p.S475L Chr.2: 86466788-86466788 on Build GRCh38
166088 c.1351_1358delAATGCACC p.N451fs*8 Chr.2: 86466715-86466722 on Build GRCh38
3963190 c.1263G>C p.K421N Chr.2: 86466627-86466627 on Build GRCh38
1632152 c.1439G>T p.R480L Chr.2: 86466803-86466803 on Build GRCh38
3963188 c.1188C>T p.N396N Chr.2: 86466552-86466552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143405154 Chr.2: 86466614-86466614 on Build GRCh38
rs756231801 Chr.2: 86466702-86466702 on Build GRCh38
rs769371668 Chr.2: 86466772-86466772 on Build GRCh38
rs200322891 Chr.2: 86466737-86466737 on Build GRCh38
rs572206443 Chr.2: 86466760-86466760 on Build GRCh38
rs377687083 Chr.2: 86466588-86466588 on Build GRCh38
rs759308577 Chr.2: 86466587-86466587 on Build GRCh38
rs1026184899 Chr.2: 86466777-86466777 on Build GRCh38
rs914451580 Chr.2: 86466719-86466719 on Build GRCh38
rs914096723 Chr.2: 86466664-86466664 on Build GRCh38
rs757341326 Chr.2: 86466591-86466591 on Build GRCh38
rs764131946 Chr.2: 86466784-86466784 on Build GRCh38
rs765006734 Chr.2: 86466692-86466692 on Build GRCh38
rs771597032 Chr.2: 86466566-86466566 on Build GRCh38
rs756390038 Chr.2: 86466600-86466600 on Build GRCh38
rs371865986 Chr.2: 86466750-86466750 on Build GRCh38
rs375633577 Chr.2: 86466646-86466646 on Build GRCh38
rs151317388 Chr.2: 86466615-86466615 on Build GRCh38
rs765972655 Chr.2: 86466576-86466576 on Build GRCh38
rs763899056 Chr.2: 86466700-86466700 on Build GRCh38
rs746573689 Chr.2: 86466743-86466743 on Build GRCh38
rs538773515 Chr.2: 86466590-86466590 on Build GRCh38
rs776239106 Chr.2: 86466676-86466676 on Build GRCh38
rs34605051 Chr.2: 86466703-86466703 on Build GRCh38
rs367637779 Chr.2: 86466741-86466741 on Build GRCh38
rs767650680 Chr.2: 86466592-86466592 on Build GRCh38
rs772389398 Chr.2: 86466654-86466654 on Build GRCh38
rs746451729 Chr.2: 86466665-86466665 on Build GRCh38
rs201238111 Chr.2: 86466682-86466682 on Build GRCh38
rs770623310 Chr.2: 86466675-86466675 on Build GRCh38
rs761294712 Chr.2: 86466562-86466562 on Build GRCh38
rs762608231 Chr.2: 86466783-86466783 on Build GRCh38
rs775092002 Chr.2: 86466693-86466693 on Build GRCh38
rs149342312 Chr.2: 86466773-86466773 on Build GRCh38
rs776325230 Chr.2: 86466759-86466759 on Build GRCh38
rs755192037 Chr.2: 86466721-86466721 on Build GRCh38
rs202001552 Chr.2: 86466659-86466659 on Build GRCh38
rs571233127 Chr.2: 86466585-86466585 on Build GRCh38
rs568759133 Chr.2: 86466627-86466627 on Build GRCh38
rs762953772 Chr.2: 86466697-86466697 on Build GRCh38
rs754097770 Chr.2: 86466704-86466704 on Build GRCh38
rs761125630 Chr.2: 86466629-86466631 on Build GRCh38
rs750404454 Chr.2: 86466599-86466599 on Build GRCh38
rs145828031 Chr.2: 86466662-86466662 on Build GRCh38
rs573499392 Chr.2: 86466701-86466701 on Build GRCh38
rs745534431 Chr.2: 86466768-86466768 on Build GRCh38
rs556530310 Chr.2: 86466603-86466603 on Build GRCh38
rs371235991 Chr.2: 86466628-86466628 on Build GRCh38
rs889167721 Chr.2: 86466705-86466705 on Build GRCh38
rs772536173 Chr.2: 86466567-86466567 on Build GRCh38
rs748418371 Chr.2: 86466733-86466733 on Build GRCh38
rs779619810 Chr.2: 86466631-86466631 on Build GRCh38
rs760376380 Chr.2: 86466568-86466568 on Build GRCh38

View Full Product Details