Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103999854-104000127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFD
Entrez Gene ID
80310
Gene Name
platelet derived growth factor D
Gene Aliases
IEGF, MSTP036, SCDGF-B, SCDGFB
UniGene
Hs.352298
Gene Chromosome Location
Chr.11: 103907186-104164299 on Build GRCh38
Transcripts
NM_025208.4 NM_033135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837681 c.319G>A p.D107N Chr.11: 104000061-104000061 on Build GRCh38
6849814 c.313G>A p.E105K Chr.11: 104000067-104000067 on Build GRCh38
5469486 c.269G>A p.R90Q Chr.11: 104000111-104000111 on Build GRCh38
3967276 c.256C>T p.Q86* Chr.11: 104000124-104000124 on Build GRCh38
1162768 c.319G>C p.D107H Chr.11: 104000061-104000061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11226108 Chr.11: 104000027-104000027 on Build GRCh38
rs191118836 Chr.11: 103999989-103999989 on Build GRCh38
rs560149055 Chr.11: 103999972-103999972 on Build GRCh38
rs746737489 Chr.11: 104000038-104000038 on Build GRCh38
rs1041920155 Chr.11: 103999904-103999904 on Build GRCh38
rs386757033 Chr.11: 103999910-103999913 on Build GRCh38
rs947553380 Chr.11: 103999889-103999889 on Build GRCh38
rs1054252419 Chr.11: 103999914-103999914 on Build GRCh38
rs200270709 Chr.11: 103999910-103999911 on Build GRCh38
rs768590440 Chr.11: 103999980-103999980 on Build GRCh38
rs1007302225 Chr.11: 103999987-103999987 on Build GRCh38
rs780980885 Chr.11: 104000083-104000083 on Build GRCh38
rs532429734 Chr.11: 103999977-103999977 on Build GRCh38
rs374742743 Chr.11: 103999897-103999901 on Build GRCh38
rs768703822 Chr.11: 104000046-104000046 on Build GRCh38
rs113541111 Chr.11: 103999911-103999911 on Build GRCh38
rs757993001 Chr.11: 104000033-104000033 on Build GRCh38
rs748312831 Chr.11: 104000099-104000099 on Build GRCh38
rs746275198 Chr.11: 103999897-103999913 on Build GRCh38
rs756698588 Chr.11: 104000026-104000026 on Build GRCh38
rs115739682 Chr.11: 103999912-103999912 on Build GRCh38
rs775534471 Chr.11: 103999877-103999877 on Build GRCh38
rs3839944 Chr.11: 104000034-104000034 on Build GRCh38
rs529909289 Chr.11: 104000079-104000079 on Build GRCh38
rs568494169 Chr.11: 103999996-103999996 on Build GRCh38
rs768604912 Chr.11: 103999896-103999896 on Build GRCh38
rs145184406 Chr.11: 103999913-103999913 on Build GRCh38
rs754922688 Chr.11: 103999909-103999911 on Build GRCh38
rs201912654 Chr.11: 103999909-103999910 on Build GRCh38
rs779834579 Chr.11: 104000035-104000035 on Build GRCh38
rs1029715988 Chr.11: 103999952-103999952 on Build GRCh38
rs951351065 Chr.11: 104000061-104000061 on Build GRCh38
rs767623763 Chr.11: 103999913-103999914 on Build GRCh38
rs974299398 Chr.11: 104000040-104000040 on Build GRCh38
rs951533019 Chr.11: 104000045-104000045 on Build GRCh38
rs891069565 Chr.11: 103999985-103999985 on Build GRCh38
rs759448274 Chr.11: 103999967-103999967 on Build GRCh38
rs753444435 Chr.11: 104000016-104000016 on Build GRCh38
rs1025498191 Chr.11: 104000057-104000057 on Build GRCh38
rs145567604 Chr.11: 103999894-103999898 on Build GRCh38
rs903416465 Chr.11: 103999906-103999906 on Build GRCh38
rs1044105517 Chr.11: 103999909-103999909 on Build GRCh38
rs753245917 Chr.11: 104000025-104000025 on Build GRCh38

View Full Product Details