Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160042303-160042562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
UniGene
Hs.408960
Gene Chromosome Location
Chr.1: 160037467-160070261 on Build GRCh38
Transcripts
NM_002241.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
282092 c.106C>T p.R36C Chr.1: 160042427-160042427 on Build GRCh38
424300 c.50G>C p.S17T Chr.1: 160042483-160042483 on Build GRCh38
7352505 c.53G>A p.R18Q Chr.1: 160042480-160042480 on Build GRCh38
4024056 c.182A>G p.D61G Chr.1: 160042351-160042351 on Build GRCh38
5787011 c.179T>C p.I60T Chr.1: 160042354-160042354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754483304 Chr.1: 160042445-160042445 on Build GRCh38
rs115466046 Chr.1: 160042480-160042480 on Build GRCh38
rs794727265 Chr.1: 160042409-160042409 on Build GRCh38
rs1130181 Chr.1: 160042384-160042384 on Build GRCh38
rs759234267 Chr.1: 160042371-160042371 on Build GRCh38
rs780870986 Chr.1: 160042451-160042451 on Build GRCh38
rs1010999260 Chr.1: 160042489-160042489 on Build GRCh38
rs776212962 Chr.1: 160042336-160042336 on Build GRCh38
rs774717149 Chr.1: 160042350-160042350 on Build GRCh38
rs200133371 Chr.1: 160042497-160042497 on Build GRCh38
rs755754492 Chr.1: 160042539-160042539 on Build GRCh38
rs72704720 Chr.1: 160042526-160042526 on Build GRCh38
rs151068619 Chr.1: 160042419-160042419 on Build GRCh38
rs776766074 Chr.1: 160042453-160042453 on Build GRCh38
rs776805483 Chr.1: 160042494-160042494 on Build GRCh38
rs773367183 Chr.1: 160042418-160042418 on Build GRCh38
rs759993423 Chr.1: 160042354-160042354 on Build GRCh38
rs779913708 Chr.1: 160042426-160042426 on Build GRCh38
rs766308095 Chr.1: 160042527-160042527 on Build GRCh38
rs1051358863 Chr.1: 160042353-160042353 on Build GRCh38
rs757578266 Chr.1: 160042392-160042392 on Build GRCh38
rs754056344 Chr.1: 160042391-160042391 on Build GRCh38
rs769464065 Chr.1: 160042454-160042454 on Build GRCh38
rs773510302 Chr.1: 160042385-160042385 on Build GRCh38
rs746292052 Chr.1: 160042470-160042470 on Build GRCh38
rs751497093 Chr.1: 160042531-160042531 on Build GRCh38
rs768340401 Chr.1: 160042323-160042323 on Build GRCh38
rs768028468 Chr.1: 160042360-160042360 on Build GRCh38
rs750246232 Chr.1: 160042400-160042400 on Build GRCh38
rs387906834 Chr.1: 160042340-160042340 on Build GRCh38
rs201907875 Chr.1: 160042390-160042390 on Build GRCh38
rs878854483 Chr.1: 160042471-160042471 on Build GRCh38
rs766560083 Chr.1: 160042450-160042450 on Build GRCh38
rs141553756 Chr.1: 160042397-160042397 on Build GRCh38
rs898999212 Chr.1: 160042416-160042416 on Build GRCh38
rs981070121 Chr.1: 160042529-160042529 on Build GRCh38
rs369062738 Chr.1: 160042456-160042456 on Build GRCh38
rs138457635 Chr.1: 160042481-160042481 on Build GRCh38
rs906427765 Chr.1: 160042468-160042468 on Build GRCh38
rs375541268 Chr.1: 160042540-160042540 on Build GRCh38
rs752204509 Chr.1: 160042537-160042537 on Build GRCh38
rs750227861 Chr.1: 160042512-160042512 on Build GRCh38
rs138943405 Chr.1: 160042457-160042457 on Build GRCh38
rs267598119 Chr.1: 160042462-160042462 on Build GRCh38
rs765568974 Chr.1: 160042510-160042510 on Build GRCh38
rs144428351 Chr.1: 160042523-160042523 on Build GRCh38
rs780525159 Chr.1: 160042535-160042535 on Build GRCh38
rs137853066 Chr.1: 160042339-160042339 on Build GRCh38
rs775905879 Chr.1: 160042479-160042479 on Build GRCh38

View Full Product Details