Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197805221-197805446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCL1
Entrez Gene ID
5334
Gene Name
phospholipase C like 1
Gene Aliases
PLCE, PLCL, PLDL1, PPP1R127, PRIP
UniGene
Hs.153322
Gene Chromosome Location
Chr.2: 197804702-198149884 on Build GRCh38
Transcripts
NM_006226.3 XM_011511351.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139164 c.146T>G p.V49G Chr.2: 197805245-197805245 on Build GRCh38
5547513 c.154delC p.P52fs*70 Chr.2: 197805253-197805253 on Build GRCh38
6364958 c.185A>G p.E62G Chr.2: 197805284-197805284 on Build GRCh38
6367747 c.197T>C p.L66P Chr.2: 197805296-197805296 on Build GRCh38
6389432 c.148G>A p.A50T Chr.2: 197805247-197805247 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs923784065 Chr.2: 197805315-197805315 on Build GRCh38
rs1051886977 Chr.2: 197805400-197805400 on Build GRCh38
rs540907120 Chr.2: 197805394-197805394 on Build GRCh38
rs949343585 Chr.2: 197805275-197805275 on Build GRCh38
rs903518697 Chr.2: 197805314-197805314 on Build GRCh38
rs991130443 Chr.2: 197805243-197805243 on Build GRCh38
rs1000964653 Chr.2: 197805319-197805319 on Build GRCh38
rs917862563 Chr.2: 197805247-197805247 on Build GRCh38
rs936384913 Chr.2: 197805356-197805356 on Build GRCh38
rs1019370030 Chr.2: 197805366-197805366 on Build GRCh38
rs747056169 Chr.2: 197805346-197805346 on Build GRCh38
rs1053505708 Chr.2: 197805364-197805364 on Build GRCh38
rs947752339 Chr.2: 197805242-197805242 on Build GRCh38
rs572200847 Chr.2: 197805392-197805392 on Build GRCh38
rs1008100034 Chr.2: 197805364-197805365 on Build GRCh38
rs765135170 Chr.2: 197805398-197805398 on Build GRCh38
rs931674523 Chr.2: 197805399-197805399 on Build GRCh38
rs558575457 Chr.2: 197805288-197805288 on Build GRCh38
rs752065199 Chr.2: 197805250-197805252 on Build GRCh38
rs757516315 Chr.2: 197805352-197805352 on Build GRCh38
rs1052350934 Chr.2: 197805338-197805338 on Build GRCh38

View Full Product Details