Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23453315-23453570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIOK3
Entrez Gene ID
8780
Gene Name
RIO kinase 3
Gene Aliases
SUDD
UniGene
Hs.445511
Gene Chromosome Location
Chr.18: 23452823-23483140 on Build GRCh38
Transcripts
NM_003831.3 XM_011526242.1 XM_011526243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750468 c.23C>T p.S8L Chr.18: 23453462-23453462 on Build GRCh38
3717841 c.26C>T p.P9L Chr.18: 23453465-23453465 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770900075 Chr.18: 23453517-23453517 on Build GRCh38
rs372168858 Chr.18: 23453535-23453535 on Build GRCh38
rs143310151 Chr.18: 23453497-23453497 on Build GRCh38
rs1052269867 Chr.18: 23453362-23453362 on Build GRCh38
rs745938445 Chr.18: 23453498-23453498 on Build GRCh38
rs758713188 Chr.18: 23453480-23453480 on Build GRCh38
rs764678492 Chr.18: 23453528-23453528 on Build GRCh38
rs545157137 Chr.18: 23453400-23453400 on Build GRCh38
rs575935869 Chr.18: 23453399-23453399 on Build GRCh38
rs756195351 Chr.18: 23453501-23453501 on Build GRCh38
rs1023301677 Chr.18: 23453419-23453419 on Build GRCh38
rs1006517985 Chr.18: 23453346-23453346 on Build GRCh38
rs764481581 Chr.18: 23453491-23453491 on Build GRCh38
rs1015965602 Chr.18: 23453355-23453355 on Build GRCh38
rs762638216 Chr.18: 23453545-23453545 on Build GRCh38
rs969692657 Chr.18: 23453361-23453361 on Build GRCh38
rs773044178 Chr.18: 23453428-23453428 on Build GRCh38
rs966993026 Chr.18: 23453531-23453531 on Build GRCh38
rs759255616 Chr.18: 23453454-23453454 on Build GRCh38
rs780185609 Chr.18: 23453348-23453348 on Build GRCh38
rs759063673 Chr.18: 23453519-23453519 on Build GRCh38
rs752290055 Chr.18: 23453472-23453472 on Build GRCh38
rs780434301 Chr.18: 23453503-23453503 on Build GRCh38
rs760617642 Chr.18: 23453437-23453437 on Build GRCh38
rs776107610 Chr.18: 23453445-23453445 on Build GRCh38
rs556171184 Chr.18: 23453383-23453383 on Build GRCh38
rs1001261897 Chr.18: 23453364-23453364 on Build GRCh38
rs993592932 Chr.18: 23453532-23453532 on Build GRCh38
rs776488572 Chr.18: 23453518-23453518 on Build GRCh38
rs764877311 Chr.18: 23453462-23453462 on Build GRCh38
rs542259930 Chr.18: 23453379-23453379 on Build GRCh38
rs1061806 Chr.18: 23453370-23453370 on Build GRCh38
rs368637057 Chr.18: 23453512-23453512 on Build GRCh38
rs896202360 Chr.18: 23453382-23453382 on Build GRCh38
rs771791165 Chr.18: 23453507-23453507 on Build GRCh38
rs374202873 Chr.18: 23453492-23453492 on Build GRCh38
rs202198326 Chr.18: 23453546-23453546 on Build GRCh38
rs757759355 Chr.18: 23453494-23453494 on Build GRCh38
rs773024712 Chr.18: 23453508-23453508 on Build GRCh38
rs749468514 Chr.18: 23453505-23453505 on Build GRCh38
rs886360761 Chr.18: 23453336-23453336 on Build GRCh38

View Full Product Details