Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51435113-51435380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC2
Entrez Gene ID
4773
Gene Name
nuclear factor of activated T-cells 2
Gene Aliases
NFAT1, NFATP
UniGene
Hs.744148
Gene Chromosome Location
Chr.20: 51386957-51562857 on Build GRCh38
Transcripts
NM_001136021.2 NM_001258292.1 NM_001258294.1 NM_001258295.1 NM_001258296.1 NM_001258297.1 NM_012340.4 NM_173091.3 XM_011528824.2 XM_011528825.2 XM_011528826.2 XM_017027850.1 XM_017027851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099400 c.1944G>T p.K648N Chr.20: 51435276-51435276 on Build GRCh38
4099398 c.1977C>T p.Y659Y Chr.20: 51435243-51435243 on Build GRCh38
6525928 c.1918G>T p.V640F Chr.20: 51435302-51435302 on Build GRCh38
7166592 c.1926C>T p.I642I Chr.20: 51435294-51435294 on Build GRCh38
6525927 c.1918G>T p.V640F Chr.20: 51435302-51435302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200465200 Chr.20: 51435168-51435168 on Build GRCh38
rs760627254 Chr.20: 51435303-51435303 on Build GRCh38
rs375392826 Chr.20: 51435169-51435169 on Build GRCh38
rs764666059 Chr.20: 51435318-51435319 on Build GRCh38
rs373087406 Chr.20: 51435277-51435277 on Build GRCh38
rs761469768 Chr.20: 51435322-51435322 on Build GRCh38
rs201762614 Chr.20: 51435351-51435351 on Build GRCh38
rs759450835 Chr.20: 51435175-51435175 on Build GRCh38
rs374852805 Chr.20: 51435344-51435344 on Build GRCh38
rs753439450 Chr.20: 51435231-51435231 on Build GRCh38
rs549354236 Chr.20: 51435198-51435198 on Build GRCh38
rs752128028 Chr.20: 51435336-51435336 on Build GRCh38
rs561066890 Chr.20: 51435242-51435242 on Build GRCh38
rs767344797 Chr.20: 51435204-51435204 on Build GRCh38
rs773542213 Chr.20: 51435284-51435284 on Build GRCh38
rs771082405 Chr.20: 51435289-51435289 on Build GRCh38
rs747362148 Chr.20: 51435144-51435144 on Build GRCh38
rs750154695 Chr.20: 51435333-51435333 on Build GRCh38
rs1042932345 Chr.20: 51435270-51435270 on Build GRCh38
rs867679254 Chr.20: 51435160-51435160 on Build GRCh38
rs528371807 Chr.20: 51435244-51435244 on Build GRCh38
rs763836299 Chr.20: 51435317-51435317 on Build GRCh38
rs952326443 Chr.20: 51435139-51435139 on Build GRCh38
rs753785304 Chr.20: 51435164-51435164 on Build GRCh38
rs766757287 Chr.20: 51435328-51435328 on Build GRCh38
rs142900177 Chr.20: 51435243-51435243 on Build GRCh38
rs766254511 Chr.20: 51435144-51435145 on Build GRCh38
rs148347040 Chr.20: 51435268-51435268 on Build GRCh38
rs775385673 Chr.20: 51435298-51435298 on Build GRCh38
rs112312123 Chr.20: 51435157-51435157 on Build GRCh38
rs148590889 Chr.20: 51435225-51435225 on Build GRCh38
rs769882225 Chr.20: 51435175-51435175 on Build GRCh38
rs756932165 Chr.20: 51435238-51435238 on Build GRCh38
rs563680481 Chr.20: 51435156-51435156 on Build GRCh38
rs778418629 Chr.20: 51435239-51435239 on Build GRCh38
rs773172999 Chr.20: 51435181-51435181 on Build GRCh38
rs111609443 Chr.20: 51435142-51435142 on Build GRCh38
rs943785272 Chr.20: 51435229-51435229 on Build GRCh38
rs752700803 Chr.20: 51435220-51435220 on Build GRCh38
rs369717696 Chr.20: 51435221-51435221 on Build GRCh38
rs151092809 Chr.20: 51435269-51435269 on Build GRCh38
rs762939847 Chr.20: 51435288-51435288 on Build GRCh38
rs769925564 Chr.20: 51435283-51435283 on Build GRCh38
rs1056285146 Chr.20: 51435326-51435326 on Build GRCh38

View Full Product Details