Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 150470995-150471254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMLD1
Entrez Gene ID
10046
Gene Name
mastermind like domain containing 1
Gene Aliases
CG1, CXorf6, F18, HYSP2
UniGene
Hs.20136
Gene Chromosome Location
Chr.X: 150361422-150514178 on Build GRCh38
Transcripts
NM_001177465.2 NM_001177466.2 NM_005491.4 XM_006724802.3 XM_006724803.3 XM_011531092.2 XM_011531093.2 XM_011531095.2 XM_017029188.1 XM_017029189.1

SNP Detail

SNP ID Chromosome Location
rs782694936 Chr.X: 150471197-150471197 on Build GRCh38
rs148402349 Chr.X: 150471019-150471019 on Build GRCh38
rs782262491 Chr.X: 150471138-150471138 on Build GRCh38
rs781802537 Chr.X: 150471205-150471205 on Build GRCh38
rs782428670 Chr.X: 150471092-150471092 on Build GRCh38
rs782589773 Chr.X: 150471117-150471117 on Build GRCh38
rs781793508 Chr.X: 150471030-150471030 on Build GRCh38
rs781854496 Chr.X: 150471232-150471232 on Build GRCh38
rs61740566 Chr.X: 150471087-150471087 on Build GRCh38
rs1006671256 Chr.X: 150471052-150471052 on Build GRCh38
rs782688486 Chr.X: 150471052-150471053 on Build GRCh38
rs371644266 Chr.X: 150471042-150471042 on Build GRCh38
rs990732997 Chr.X: 150471101-150471101 on Build GRCh38
rs782655809 Chr.X: 150471084-150471084 on Build GRCh38
rs782768625 Chr.X: 150471177-150471177 on Build GRCh38
rs371252489 Chr.X: 150471190-150471190 on Build GRCh38
rs145699612 Chr.X: 150471146-150471146 on Build GRCh38
rs142352628 Chr.X: 150471129-150471129 on Build GRCh38
rs782090608 Chr.X: 150471193-150471193 on Build GRCh38
rs782520899 Chr.X: 150471064-150471065 on Build GRCh38
rs782267505 Chr.X: 150471125-150471125 on Build GRCh38
rs367735248 Chr.X: 150471081-150471081 on Build GRCh38
rs782017313 Chr.X: 150471170-150471170 on Build GRCh38
rs782538359 Chr.X: 150471212-150471212 on Build GRCh38
rs781915260 Chr.X: 150471145-150471145 on Build GRCh38
rs146302921 Chr.X: 150471130-150471130 on Build GRCh38
rs782668343 Chr.X: 150471224-150471224 on Build GRCh38
rs61739630 Chr.X: 150471112-150471112 on Build GRCh38
rs782160809 Chr.X: 150471175-150471175 on Build GRCh38
rs782076071 Chr.X: 150471016-150471016 on Build GRCh38
rs782324407 Chr.X: 150471153-150471153 on Build GRCh38

View Full Product Details