Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55401233-55401502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6C65
Entrez Gene ID
403282
Gene Name
olfactory receptor family 6 subfamily C member 65
Gene Aliases
-
UniGene
Hs.554553
Gene Chromosome Location
Chr.12: 55400529-55401467 on Build GRCh38
Transcripts
NM_001005518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399127 c.853C>T p.P285S Chr.12: 55401381-55401381 on Build GRCh38
6462724 c.939A>G p.*313* Chr.12: 55401467-55401467 on Build GRCh38
941292 c.731T>G p.I244S Chr.12: 55401259-55401259 on Build GRCh38
941293 c.762T>G p.I254M Chr.12: 55401290-55401290 on Build GRCh38
4043288 c.888A>G p.K296K Chr.12: 55401416-55401416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779756982 Chr.12: 55401366-55401366 on Build GRCh38
rs143868119 Chr.12: 55401416-55401416 on Build GRCh38
rs913878450 Chr.12: 55401352-55401352 on Build GRCh38
rs760145668 Chr.12: 55401311-55401311 on Build GRCh38
rs757254424 Chr.12: 55401341-55401341 on Build GRCh38
rs867602186 Chr.12: 55401346-55401346 on Build GRCh38
rs764561279 Chr.12: 55401421-55401421 on Build GRCh38
rs751875427 Chr.12: 55401327-55401327 on Build GRCh38
rs770634906 Chr.12: 55401265-55401266 on Build GRCh38
rs749095229 Chr.12: 55401367-55401367 on Build GRCh38
rs61744496 Chr.12: 55401447-55401447 on Build GRCh38
rs777029838 Chr.12: 55401410-55401410 on Build GRCh38
rs775288325 Chr.12: 55401445-55401445 on Build GRCh38
rs778422272 Chr.12: 55401381-55401381 on Build GRCh38
rs981047605 Chr.12: 55401308-55401308 on Build GRCh38
rs530999009 Chr.12: 55401394-55401394 on Build GRCh38
rs750309229 Chr.12: 55401344-55401344 on Build GRCh38
rs769246593 Chr.12: 55401325-55401325 on Build GRCh38
rs771457969 Chr.12: 55401383-55401383 on Build GRCh38
rs771449948 Chr.12: 55401315-55401315 on Build GRCh38
rs755988092 Chr.12: 55401462-55401462 on Build GRCh38
rs372659272 Chr.12: 55401388-55401388 on Build GRCh38
rs202236592 Chr.12: 55401283-55401283 on Build GRCh38
rs771932197 Chr.12: 55401437-55401438 on Build GRCh38
rs770441022 Chr.12: 55401395-55401395 on Build GRCh38
rs776201382 Chr.12: 55401397-55401397 on Build GRCh38
rs756076045 Chr.12: 55401361-55401361 on Build GRCh38
rs776970420 Chr.12: 55401294-55401294 on Build GRCh38
rs774416422 Chr.12: 55401444-55401444 on Build GRCh38
rs267603567 Chr.12: 55401318-55401318 on Build GRCh38
rs768550494 Chr.12: 55401373-55401373 on Build GRCh38
rs767454895 Chr.12: 55401455-55401455 on Build GRCh38
rs750504955 Chr.12: 55401460-55401460 on Build GRCh38
rs754694119 Chr.12: 55401476-55401476 on Build GRCh38
rs945400047 Chr.12: 55401353-55401353 on Build GRCh38
rs745768296 Chr.12: 55401341-55401361 on Build GRCh38
rs778685978 Chr.12: 55401294-55401294 on Build GRCh38
rs932387115 Chr.12: 55401398-55401398 on Build GRCh38
rs760987828 Chr.12: 55401261-55401261 on Build GRCh38
rs781383486 Chr.12: 55401343-55401343 on Build GRCh38
rs765660126 Chr.12: 55401314-55401314 on Build GRCh38
rs138141433 Chr.12: 55401359-55401359 on Build GRCh38
rs761212603 Chr.12: 55401280-55401280 on Build GRCh38
rs139289386 Chr.12: 55401340-55401340 on Build GRCh38
rs1028229472 Chr.12: 55401278-55401278 on Build GRCh38
rs753835499 Chr.12: 55401471-55401471 on Build GRCh38

View Full Product Details