Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196488534-196488761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF168
Entrez Gene ID
165918
Gene Name
ring finger protein 168
Gene Aliases
hRNF168
UniGene
Hs.250648
Gene Chromosome Location
Chr.3: 196468783-196503768 on Build GRCh38
Transcripts
NM_152617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1422004 c.323G>A p.R108H Chr.3: 196488662-196488662 on Build GRCh38
1043041 c.378+1G>A p.? Chr.3: 196488606-196488606 on Build GRCh38
270049 c.336A>C p.K112N Chr.3: 196488649-196488649 on Build GRCh38
3591868 c.353G>A p.R118K Chr.3: 196488632-196488632 on Build GRCh38
3591869 c.345A>G p.E115E Chr.3: 196488640-196488640 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567216760 Chr.3: 196488581-196488582 on Build GRCh38
rs759194563 Chr.3: 196488635-196488635 on Build GRCh38
rs144091604 Chr.3: 196488683-196488683 on Build GRCh38
rs755261571 Chr.3: 196488674-196488674 on Build GRCh38
rs533981052 Chr.3: 196488582-196488583 on Build GRCh38
rs750986787 Chr.3: 196488658-196488658 on Build GRCh38
rs775164430 Chr.3: 196488712-196488712 on Build GRCh38
rs569352140 Chr.3: 196488582-196488582 on Build GRCh38
rs756026515 Chr.3: 196488680-196488680 on Build GRCh38
rs772733440 Chr.3: 196488588-196488588 on Build GRCh38
rs781135811 Chr.3: 196488581-196488581 on Build GRCh38
rs747312129 Chr.3: 196488577-196488577 on Build GRCh38
rs557320846 Chr.3: 196488717-196488717 on Build GRCh38
rs748038395 Chr.3: 196488582-196488582 on Build GRCh38
rs377631557 Chr.3: 196488692-196488692 on Build GRCh38
rs149773824 Chr.3: 196488678-196488678 on Build GRCh38
rs751761904 Chr.3: 196488669-196488669 on Build GRCh38
rs763609874 Chr.3: 196488608-196488608 on Build GRCh38
rs771651426 Chr.3: 196488617-196488619 on Build GRCh38
rs1026977947 Chr.3: 196488591-196488591 on Build GRCh38
rs749317182 Chr.3: 196488685-196488685 on Build GRCh38
rs371040643 Chr.3: 196488706-196488706 on Build GRCh38
rs192270033 Chr.3: 196488732-196488732 on Build GRCh38
rs62798260 Chr.3: 196488581-196488581 on Build GRCh38
rs978802446 Chr.3: 196488687-196488687 on Build GRCh38
rs754813213 Chr.3: 196488580-196488580 on Build GRCh38
rs201800722 Chr.3: 196488597-196488597 on Build GRCh38
rs777000132 Chr.3: 196488579-196488579 on Build GRCh38
rs765630387 Chr.3: 196488655-196488655 on Build GRCh38
rs770297464 Chr.3: 196488580-196488581 on Build GRCh38
rs993713391 Chr.3: 196488578-196488578 on Build GRCh38
rs369774165 Chr.3: 196488724-196488724 on Build GRCh38
rs769871467 Chr.3: 196488585-196488585 on Build GRCh38
rs778500554 Chr.3: 196488700-196488700 on Build GRCh38
rs764386439 Chr.3: 196488652-196488652 on Build GRCh38
rs114080378 Chr.3: 196488713-196488713 on Build GRCh38
rs373932426 Chr.3: 196488593-196488593 on Build GRCh38
rs528924586 Chr.3: 196488654-196488654 on Build GRCh38
rs112381143 Chr.3: 196488569-196488569 on Build GRCh38
rs745475922 Chr.3: 196488702-196488702 on Build GRCh38
rs1009232382 Chr.3: 196488719-196488719 on Build GRCh38
rs781082241 Chr.3: 196488677-196488677 on Build GRCh38
rs766545137 Chr.3: 196488662-196488662 on Build GRCh38
rs748729157 Chr.3: 196488590-196488590 on Build GRCh38
rs773872938 Chr.3: 196488602-196488602 on Build GRCh38
rs893542447 Chr.3: 196488690-196488690 on Build GRCh38
rs376982185 Chr.3: 196488715-196488715 on Build GRCh38
rs762927567 Chr.3: 196488661-196488661 on Build GRCh38
rs774573354 Chr.3: 196488627-196488627 on Build GRCh38
rs766309035 Chr.3: 196488731-196488731 on Build GRCh38

View Full Product Details