Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129639305-129639538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3904718 c.356T>C p.V119A Chr.9: 129639448-129639448 on Build GRCh38
3926280 c.398A>G p.D133G Chr.9: 129639406-129639406 on Build GRCh38
1105868 c.392G>A p.R131Q Chr.9: 129639412-129639412 on Build GRCh38
4407435 c.327C>G p.I109M Chr.9: 129639477-129639477 on Build GRCh38
7256027 c.375C>T p.H125H Chr.9: 129639429-129639429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144505727 Chr.9: 129639490-129639490 on Build GRCh38
rs41278724 Chr.9: 129639426-129639426 on Build GRCh38
rs762336302 Chr.9: 129639388-129639388 on Build GRCh38
rs149231958 Chr.9: 129639478-129639478 on Build GRCh38
rs752452958 Chr.9: 129639410-129639410 on Build GRCh38
rs750436777 Chr.9: 129639514-129639514 on Build GRCh38
rs767827766 Chr.9: 129639508-129639508 on Build GRCh38
rs377001543 Chr.9: 129639489-129639489 on Build GRCh38
rs943655371 Chr.9: 129639384-129639384 on Build GRCh38
rs776601015 Chr.9: 129639328-129639328 on Build GRCh38
rs977203458 Chr.9: 129639334-129639334 on Build GRCh38
rs781128641 Chr.9: 129639428-129639428 on Build GRCh38
rs766648586 Chr.9: 129639408-129639408 on Build GRCh38
rs770962165 Chr.9: 129639327-129639327 on Build GRCh38
rs758101815 Chr.9: 129639412-129639412 on Build GRCh38
rs145778904 Chr.9: 129639403-129639403 on Build GRCh38
rs772570376 Chr.9: 129639471-129639471 on Build GRCh38
rs935684179 Chr.9: 129639343-129639343 on Build GRCh38
rs745629378 Chr.9: 129639333-129639333 on Build GRCh38
rs377139298 Chr.9: 129639517-129639517 on Build GRCh38
rs774433494 Chr.9: 129639457-129639457 on Build GRCh38
rs61758721 Chr.9: 129639436-129639436 on Build GRCh38
rs370196393 Chr.9: 129639479-129639479 on Build GRCh38
rs537770792 Chr.9: 129639477-129639477 on Build GRCh38
rs745455005 Chr.9: 129639386-129639386 on Build GRCh38
rs749561289 Chr.9: 129639452-129639452 on Build GRCh38
rs971227545 Chr.9: 129639496-129639496 on Build GRCh38
rs375966860 Chr.9: 129639423-129639423 on Build GRCh38
rs767960913 Chr.9: 129639401-129639401 on Build GRCh38
rs138385332 Chr.9: 129639473-129639473 on Build GRCh38
rs547799738 Chr.9: 129639501-129639501 on Build GRCh38
rs745657553 Chr.9: 129639432-129639432 on Build GRCh38
rs144625798 Chr.9: 129639456-129639456 on Build GRCh38
rs374379078 Chr.9: 129639372-129639372 on Build GRCh38
rs1133536 Chr.9: 129639488-129639488 on Build GRCh38
rs371148084 Chr.9: 129639340-129639340 on Build GRCh38
rs144137235 Chr.9: 129639474-129639474 on Build GRCh38
rs775717676 Chr.9: 129639344-129639344 on Build GRCh38
rs763005968 Chr.9: 129639350-129639350 on Build GRCh38
rs138255998 Chr.9: 129639363-129639363 on Build GRCh38
rs777467438 Chr.9: 129639422-129639422 on Build GRCh38
rs769302619 Chr.9: 129639419-129639421 on Build GRCh38
rs369808920 Chr.9: 129639364-129639364 on Build GRCh38
rs761032374 Chr.9: 129639404-129639404 on Build GRCh38
rs779927342 Chr.9: 129639518-129639518 on Build GRCh38
rs889389967 Chr.9: 129639370-129639370 on Build GRCh38
rs1026364095 Chr.9: 129639468-129639468 on Build GRCh38
rs764202828 Chr.9: 129639354-129639354 on Build GRCh38
rs766816681 Chr.9: 129639476-129639476 on Build GRCh38
rs367980061 Chr.9: 129639450-129639450 on Build GRCh38
rs543132075 Chr.9: 129639466-129639466 on Build GRCh38

View Full Product Details