Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65918031-65918284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf68
Entrez Gene ID
83638
Gene Name
chromosome 11 open reading frame 68
Gene Aliases
BLES03, P5326
UniGene
Hs.433573
Gene Chromosome Location
Chr.11: 65916810-65919060 on Build GRCh38
Transcripts
NM_001135635.1 NM_031450.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2166810 c.127A>T p.M43L Chr.11: 65918214-65918214 on Build GRCh38
5498027 c.62C>G p.T21S Chr.11: 65918153-65918153 on Build GRCh38
7002154 c.143_145AGC>18 p.E48>GPGFIP Chr.11: 65918196-65918198 on Build GRCh38
2166811 c.1A>T p.M1L Chr.11: 65918214-65918214 on Build GRCh38
144101 c.125G>A p.R42K Chr.11: 65918216-65918216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375631477 Chr.11: 65918221-65918221 on Build GRCh38
rs778946994 Chr.11: 65918137-65918137 on Build GRCh38
rs762690740 Chr.11: 65918159-65918159 on Build GRCh38
rs35868735 Chr.11: 65918174-65918174 on Build GRCh38
rs961495072 Chr.11: 65918214-65918214 on Build GRCh38
rs781019646 Chr.11: 65918148-65918148 on Build GRCh38
rs769726744 Chr.11: 65918152-65918152 on Build GRCh38
rs755258694 Chr.11: 65918131-65918131 on Build GRCh38
rs768480845 Chr.11: 65918182-65918182 on Build GRCh38
rs748221899 Chr.11: 65918140-65918140 on Build GRCh38
rs775307507 Chr.11: 65918080-65918080 on Build GRCh38
rs754063393 Chr.11: 65918114-65918114 on Build GRCh38
rs770867726 Chr.11: 65918143-65918143 on Build GRCh38
rs955315431 Chr.11: 65918179-65918179 on Build GRCh38
rs745631066 Chr.11: 65918149-65918149 on Build GRCh38
rs754255448 Chr.11: 65918236-65918236 on Build GRCh38
rs191917397 Chr.11: 65918108-65918108 on Build GRCh38
rs572025542 Chr.11: 65918185-65918185 on Build GRCh38
rs779731203 Chr.11: 65918162-65918185 on Build GRCh38
rs73496433 Chr.11: 65918255-65918255 on Build GRCh38
rs750770390 Chr.11: 65918090-65918090 on Build GRCh38
rs750905797 Chr.11: 65918224-65918224 on Build GRCh38
rs761032572 Chr.11: 65918226-65918226 on Build GRCh38
rs985069815 Chr.11: 65918253-65918253 on Build GRCh38
rs1025362061 Chr.11: 65918088-65918088 on Build GRCh38
rs565007272 Chr.11: 65918110-65918110 on Build GRCh38
rs762368596 Chr.11: 65918086-65918086 on Build GRCh38
rs776462496 Chr.11: 65918052-65918052 on Build GRCh38
rs767960379 Chr.11: 65918087-65918087 on Build GRCh38
rs759287565 Chr.11: 65918057-65918057 on Build GRCh38
rs765481469 Chr.11: 65918251-65918251 on Build GRCh38
rs763705751 Chr.11: 65918081-65918081 on Build GRCh38
rs773978540 Chr.11: 65918190-65918190 on Build GRCh38
rs771807885 Chr.11: 65918212-65918212 on Build GRCh38
rs775219397 Chr.11: 65918158-65918158 on Build GRCh38
rs368300054 Chr.11: 65918232-65918232 on Build GRCh38
rs755166518 Chr.11: 65918249-65918249 on Build GRCh38
rs769451839 Chr.11: 65918064-65918064 on Build GRCh38
rs925199702 Chr.11: 65918244-65918244 on Build GRCh38
rs868288399 Chr.11: 65918156-65918156 on Build GRCh38
rs751182740 Chr.11: 65918082-65918082 on Build GRCh38

View Full Product Details