Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110650365-110650626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960967 c.2203+5G>T p.? Chr.2: 110650541-110650541 on Build GRCh38
5772621 c.2203+4C>T p.? Chr.2: 110650542-110650542 on Build GRCh38
6355844 c.2159C>T p.A720V Chr.2: 110650590-110650590 on Build GRCh38
1613288 c.2187G>T p.G729G Chr.2: 110650562-110650562 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751876699 Chr.2: 110650596-110650596 on Build GRCh38
rs755350549 Chr.2: 110650597-110650597 on Build GRCh38
rs768662095 Chr.2: 110650566-110650566 on Build GRCh38
rs577463953 Chr.2: 110650591-110650591 on Build GRCh38
rs780263145 Chr.2: 110650501-110650501 on Build GRCh38
rs999056328 Chr.2: 110650402-110650402 on Build GRCh38
rs773393098 Chr.2: 110650592-110650592 on Build GRCh38
rs776719073 Chr.2: 110650567-110650567 on Build GRCh38
rs868578212 Chr.2: 110650497-110650497 on Build GRCh38
rs373297870 Chr.2: 110650542-110650542 on Build GRCh38
rs1045594666 Chr.2: 110650484-110650484 on Build GRCh38
rs763057015 Chr.2: 110650593-110650593 on Build GRCh38
rs766659533 Chr.2: 110650594-110650594 on Build GRCh38
rs748738807 Chr.2: 110650539-110650539 on Build GRCh38
rs949547482 Chr.2: 110650467-110650467 on Build GRCh38
rs893308096 Chr.2: 110650512-110650512 on Build GRCh38
rs560957245 Chr.2: 110650403-110650403 on Build GRCh38
rs761914583 Chr.2: 110650571-110650571 on Build GRCh38
rs147498621 Chr.2: 110650390-110650390 on Build GRCh38
rs760577088 Chr.2: 110650564-110650564 on Build GRCh38
rs892450555 Chr.2: 110650552-110650552 on Build GRCh38
rs972249054 Chr.2: 110650453-110650453 on Build GRCh38
rs905256346 Chr.2: 110650494-110650494 on Build GRCh38
rs771803937 Chr.2: 110650551-110650551 on Build GRCh38
rs751962820 Chr.2: 110650509-110650509 on Build GRCh38
rs1023062528 Chr.2: 110650555-110650555 on Build GRCh38
rs918093361 Chr.2: 110650458-110650458 on Build GRCh38
rs778553647 Chr.2: 110650546-110650546 on Build GRCh38
rs557116866 Chr.2: 110650562-110650562 on Build GRCh38
rs940964655 Chr.2: 110650396-110650396 on Build GRCh38
rs781683255 Chr.2: 110650526-110650526 on Build GRCh38
rs543760205 Chr.2: 110650549-110650549 on Build GRCh38
rs755442449 Chr.2: 110650522-110650522 on Build GRCh38

View Full Product Details