Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110251271-110251544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXN
Entrez Gene ID
7295
Gene Name
thioredoxin
Gene Aliases
TRDX, TRX, TRX1
UniGene
Hs.435136
Gene Chromosome Location
Chr.9: 110243812-110256640 on Build GRCh38
Transcripts
NM_001244938.1 NM_003329.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654414 c.33T>G p.F11L Chr.9: 110251454-110251454 on Build GRCh38
6292840 c.114C>A p.I38I Chr.9: 110251373-110251373 on Build GRCh38
1103820 c.81C>T p.F27F Chr.9: 110251406-110251406 on Build GRCh38
4510511 c.83C>G p.S28* Chr.9: 110251404-110251404 on Build GRCh38
3996226 c.72A>G p.V24V Chr.9: 110251415-110251415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904185868 Chr.9: 110251469-110251469 on Build GRCh38
rs952113585 Chr.9: 110251414-110251414 on Build GRCh38
rs761633708 Chr.9: 110251504-110251504 on Build GRCh38
rs767171958 Chr.9: 110251361-110251361 on Build GRCh38
rs752247588 Chr.9: 110251367-110251367 on Build GRCh38
rs766604635 Chr.9: 110251489-110251489 on Build GRCh38
rs4135192 Chr.9: 110251308-110251308 on Build GRCh38
rs756707534 Chr.9: 110251312-110251313 on Build GRCh38
rs145669620 Chr.9: 110251383-110251383 on Build GRCh38
rs533670835 Chr.9: 110251454-110251454 on Build GRCh38
rs760750545 Chr.9: 110251371-110251371 on Build GRCh38
rs375581079 Chr.9: 110251365-110251365 on Build GRCh38
rs369965207 Chr.9: 110251421-110251421 on Build GRCh38
rs998641470 Chr.9: 110251482-110251482 on Build GRCh38
rs749648542 Chr.9: 110251426-110251426 on Build GRCh38
rs12047 Chr.9: 110251386-110251386 on Build GRCh38
rs897689772 Chr.9: 110251305-110251305 on Build GRCh38
rs889012440 Chr.9: 110251420-110251420 on Build GRCh38
rs745670693 Chr.9: 110251298-110251298 on Build GRCh38
rs780710024 Chr.9: 110251488-110251491 on Build GRCh38
rs781316635 Chr.9: 110251403-110251403 on Build GRCh38
rs763968382 Chr.9: 110251462-110251462 on Build GRCh38
rs565594763 Chr.9: 110251399-110251399 on Build GRCh38
rs774019220 Chr.9: 110251355-110251355 on Build GRCh38
rs757034945 Chr.9: 110251389-110251389 on Build GRCh38
rs933119715 Chr.9: 110251391-110251391 on Build GRCh38
rs773508882 Chr.9: 110251332-110251332 on Build GRCh38
rs774562370 Chr.9: 110251433-110251433 on Build GRCh38
rs763320812 Chr.9: 110251335-110251335 on Build GRCh38
rs770497055 Chr.9: 110251348-110251348 on Build GRCh38
rs1012933057 Chr.9: 110251415-110251415 on Build GRCh38
rs200412877 Chr.9: 110251397-110251397 on Build GRCh38
rs776581885 Chr.9: 110251477-110251477 on Build GRCh38
rs201616240 Chr.9: 110251315-110251315 on Build GRCh38
rs373394120 Chr.9: 110251444-110251444 on Build GRCh38
rs138828481 Chr.9: 110251439-110251439 on Build GRCh38
rs765120372 Chr.9: 110251513-110251513 on Build GRCh38
rs547387406 Chr.9: 110251398-110251398 on Build GRCh38
rs146018216 Chr.9: 110251437-110251437 on Build GRCh38
rs764062295 Chr.9: 110251373-110251373 on Build GRCh38
rs781227350 Chr.9: 110251320-110251320 on Build GRCh38
rs758994917 Chr.9: 110251358-110251358 on Build GRCh38
rs748213654 Chr.9: 110251406-110251406 on Build GRCh38
rs748693917 Chr.9: 110251325-110251325 on Build GRCh38
rs373433889 Chr.9: 110251438-110251438 on Build GRCh38
rs142050233 Chr.9: 110251388-110251388 on Build GRCh38
rs771282320 Chr.9: 110251429-110251429 on Build GRCh38
rs770300484 Chr.9: 110251418-110251418 on Build GRCh38
rs909861031 Chr.9: 110251339-110251339 on Build GRCh38
rs997392 Chr.9: 110251326-110251326 on Build GRCh38

View Full Product Details