Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3005530-3005769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE2
Entrez Gene ID
7089
Gene Name
transducin like enhancer of split 2
Gene Aliases
ESG, ESG2, GRG2
UniGene
Hs.332173
Gene Chromosome Location
Chr.19: 2997638-3047635 on Build GRCh38
Transcripts
NM_001144761.1 NM_001144762.1 NM_001300846.1 NM_003260.4 XM_006722864.2 XM_011528230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201126 c.1748+4G>C p.? Chr.19: 3005717-3005717 on Build GRCh38
6902286 c.1717G>A p.V573I Chr.19: 3005752-3005752 on Build GRCh38
4856861 c.1793C>T p.S598F Chr.19: 3005540-3005540 on Build GRCh38
7202006 c.1795G>A p.D599N Chr.19: 3005538-3005538 on Build GRCh38
6802201 c.1780T>C p.C594R Chr.19: 3005553-3005553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539545418 Chr.19: 3005606-3005606 on Build GRCh38
rs1009188313 Chr.19: 3005687-3005687 on Build GRCh38
rs374951690 Chr.19: 3005710-3005710 on Build GRCh38
rs181337463 Chr.19: 3005691-3005691 on Build GRCh38
rs557761365 Chr.19: 3005654-3005654 on Build GRCh38
rs757384030 Chr.19: 3005634-3005634 on Build GRCh38
rs749556181 Chr.19: 3005623-3005623 on Build GRCh38
rs750098452 Chr.19: 3005562-3005562 on Build GRCh38
rs762407936 Chr.19: 3005563-3005563 on Build GRCh38
rs754925988 Chr.19: 3005588-3005588 on Build GRCh38
rs778096364 Chr.19: 3005616-3005616 on Build GRCh38
rs200595351 Chr.19: 3005685-3005685 on Build GRCh38
rs368477804 Chr.19: 3005675-3005675 on Build GRCh38
rs776077156 Chr.19: 3005682-3005682 on Build GRCh38
rs762639964 Chr.19: 3005705-3005705 on Build GRCh38
rs755936856 Chr.19: 3005739-3005739 on Build GRCh38
rs367942728 Chr.19: 3005750-3005750 on Build GRCh38
rs772521808 Chr.19: 3005680-3005680 on Build GRCh38
rs868048027 Chr.19: 3005662-3005662 on Build GRCh38
rs374442281 Chr.19: 3005566-3005566 on Build GRCh38
rs34703558 Chr.19: 3005557-3005557 on Build GRCh38
rs767303661 Chr.19: 3005728-3005728 on Build GRCh38
rs752704765 Chr.19: 3005730-3005730 on Build GRCh38
rs982586741 Chr.19: 3005567-3005567 on Build GRCh38
rs773795282 Chr.19: 3005706-3005706 on Build GRCh38
rs891620849 Chr.19: 3005560-3005560 on Build GRCh38
rs780554324 Chr.19: 3005660-3005660 on Build GRCh38
rs746375322 Chr.19: 3005679-3005679 on Build GRCh38
rs751490192 Chr.19: 3005577-3005577 on Build GRCh38
rs752364795 Chr.19: 3005597-3005597 on Build GRCh38
rs565878660 Chr.19: 3005591-3005591 on Build GRCh38

View Full Product Details