Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6872702-6872973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPSB2
Entrez Gene ID
84727
Gene Name
splA/ryanodine receptor domain and SOCS box containing 2
Gene Aliases
GRCC9, SSB2
UniGene
Hs.479856
Gene Chromosome Location
Chr.12: 6870935-6873357 on Build GRCh38
Transcripts
NM_001146316.1 NM_001319670.1 NM_032641.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173579 c.69C>G p.L23L Chr.12: 6872833-6872833 on Build GRCh38
4968088 c.72C>A p.S24S Chr.12: 6872830-6872830 on Build GRCh38
5818063 c.128A>T p.Q43L Chr.12: 6872774-6872774 on Build GRCh38
6789389 c.107C>A p.P36H Chr.12: 6872795-6872795 on Build GRCh38
4044510 c.152A>G p.K51R Chr.12: 6872750-6872750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782466615 Chr.12: 6872917-6872917 on Build GRCh38
rs141265707 Chr.12: 6872781-6872781 on Build GRCh38
rs782382559 Chr.12: 6872933-6872933 on Build GRCh38
rs782629108 Chr.12: 6872919-6872919 on Build GRCh38
rs782290323 Chr.12: 6872822-6872822 on Build GRCh38
rs782041206 Chr.12: 6872880-6872880 on Build GRCh38
rs376409580 Chr.12: 6872908-6872908 on Build GRCh38
rs1015854542 Chr.12: 6872779-6872779 on Build GRCh38
rs782004250 Chr.12: 6872941-6872941 on Build GRCh38
rs923020537 Chr.12: 6872828-6872828 on Build GRCh38
rs781896519 Chr.12: 6872882-6872882 on Build GRCh38
rs181213322 Chr.12: 6872826-6872826 on Build GRCh38
rs782557308 Chr.12: 6872736-6872736 on Build GRCh38
rs782407884 Chr.12: 6872866-6872866 on Build GRCh38
rs189782769 Chr.12: 6872778-6872778 on Build GRCh38
rs782102272 Chr.12: 6872790-6872790 on Build GRCh38
rs985746433 Chr.12: 6872927-6872927 on Build GRCh38
rs782363158 Chr.12: 6872847-6872847 on Build GRCh38
rs782449386 Chr.12: 6872813-6872813 on Build GRCh38
rs782302342 Chr.12: 6872929-6872929 on Build GRCh38
rs782473254 Chr.12: 6872792-6872792 on Build GRCh38
rs782281673 Chr.12: 6872725-6872725 on Build GRCh38
rs782806744 Chr.12: 6872793-6872793 on Build GRCh38
rs782553519 Chr.12: 6872910-6872910 on Build GRCh38
rs782527289 Chr.12: 6872924-6872924 on Build GRCh38
rs782176780 Chr.12: 6872801-6872801 on Build GRCh38
rs782640274 Chr.12: 6872737-6872737 on Build GRCh38
rs781827108 Chr.12: 6872763-6872763 on Build GRCh38
rs138023519 Chr.12: 6872858-6872858 on Build GRCh38
rs782211445 Chr.12: 6872740-6872740 on Build GRCh38
rs781786881 Chr.12: 6872785-6872785 on Build GRCh38
rs782327945 Chr.12: 6872921-6872921 on Build GRCh38
rs781828555 Chr.12: 6872928-6872928 on Build GRCh38
rs782566062 Chr.12: 6872817-6872817 on Build GRCh38
rs782622112 Chr.12: 6872913-6872913 on Build GRCh38
rs782045508 Chr.12: 6872896-6872896 on Build GRCh38
rs782215852 Chr.12: 6872831-6872831 on Build GRCh38
rs200000609 Chr.12: 6872728-6872728 on Build GRCh38
rs1027012630 Chr.12: 6872748-6872748 on Build GRCh38
rs781914112 Chr.12: 6872749-6872749 on Build GRCh38
rs781922227 Chr.12: 6872859-6872859 on Build GRCh38
rs781833150 Chr.12: 6872895-6872895 on Build GRCh38
rs782030794 Chr.12: 6872769-6872769 on Build GRCh38
rs545526842 Chr.12: 6872902-6872902 on Build GRCh38
rs782672712 Chr.12: 6872820-6872820 on Build GRCh38
rs782708793 Chr.12: 6872791-6872791 on Build GRCh38
rs782798040 Chr.12: 6872889-6872889 on Build GRCh38
rs782353887 Chr.12: 6872744-6872744 on Build GRCh38
rs781868918 Chr.12: 6872914-6872914 on Build GRCh38
rs782612726 Chr.12: 6872794-6872794 on Build GRCh38
rs782068809 Chr.12: 6872864-6872864 on Build GRCh38
rs61733175 Chr.12: 6872827-6872827 on Build GRCh38
rs782192458 Chr.12: 6872752-6872752 on Build GRCh38

View Full Product Details