Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55720435-55720696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM100
Entrez Gene ID
55273
Gene Name
transmembrane protein 100
Gene Aliases
-
UniGene
Hs.173233
Gene Chromosome Location
Chr.17: 55719627-55732121 on Build GRCh38
Transcripts
NM_001099640.1 NM_018286.2 XM_017024814.1 XM_017024815.1 XM_017024816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983889 c.380C>A p.A127E Chr.17: 55720691-55720691 on Build GRCh38
7152759 c.375C>G p.L125L Chr.17: 55720696-55720696 on Build GRCh38
6803054 c.394T>G p.L132V Chr.17: 55720677-55720677 on Build GRCh38
318120 c.401C>A p.A134D Chr.17: 55720670-55720670 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770174759 Chr.17: 55720670-55720670 on Build GRCh38
rs1050086850 Chr.17: 55720611-55720611 on Build GRCh38
rs1023273124 Chr.17: 55720513-55720513 on Build GRCh38
rs77955953 Chr.17: 55720588-55720588 on Build GRCh38
rs774070404 Chr.17: 55720584-55720584 on Build GRCh38
rs9893242 Chr.17: 55720632-55720632 on Build GRCh38
rs185469303 Chr.17: 55720567-55720567 on Build GRCh38
rs560648250 Chr.17: 55720542-55720542 on Build GRCh38
rs575200659 Chr.17: 55720502-55720502 on Build GRCh38
rs970016008 Chr.17: 55720520-55720520 on Build GRCh38
rs972789963 Chr.17: 55720558-55720558 on Build GRCh38
rs775168360 Chr.17: 55720487-55720487 on Build GRCh38
rs746054735 Chr.17: 55720642-55720642 on Build GRCh38
rs890013884 Chr.17: 55720627-55720627 on Build GRCh38
rs960123796 Chr.17: 55720492-55720492 on Build GRCh38
rs780229217 Chr.17: 55720654-55720654 on Build GRCh38
rs768982336 Chr.17: 55720657-55720657 on Build GRCh38
rs3803827 Chr.17: 55720547-55720547 on Build GRCh38
rs763453197 Chr.17: 55720623-55720643 on Build GRCh38
rs148265718 Chr.17: 55720668-55720668 on Build GRCh38
rs1049939050 Chr.17: 55720597-55720597 on Build GRCh38
rs531835496 Chr.17: 55720568-55720568 on Build GRCh38
rs201854868 Chr.17: 55720648-55720648 on Build GRCh38
rs778620404 Chr.17: 55720629-55720629 on Build GRCh38
rs376662097 Chr.17: 55720655-55720655 on Build GRCh38
rs907698415 Chr.17: 55720573-55720573 on Build GRCh38
rs992701715 Chr.17: 55720510-55720510 on Build GRCh38
rs571097892 Chr.17: 55720590-55720590 on Build GRCh38
rs773608196 Chr.17: 55720675-55720675 on Build GRCh38
rs748248873 Chr.17: 55720605-55720605 on Build GRCh38
rs747094386 Chr.17: 55720656-55720656 on Build GRCh38
rs946957580 Chr.17: 55720641-55720641 on Build GRCh38
rs753784060 Chr.17: 55720625-55720625 on Build GRCh38
rs984047856 Chr.17: 55720524-55720524 on Build GRCh38
rs181291697 Chr.17: 55720543-55720543 on Build GRCh38
rs777194276 Chr.17: 55720659-55720659 on Build GRCh38
rs757059935 Chr.17: 55720626-55720626 on Build GRCh38
rs372576404 Chr.17: 55720637-55720637 on Build GRCh38
rs552803237 Chr.17: 55720534-55720534 on Build GRCh38

View Full Product Details