Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8841310-8841583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
UniGene
Hs.620532
Gene Chromosome Location
Chr.12: 8822472-8887202 on Build GRCh38
Transcripts
NM_144670.5 XM_011520566.2 XM_011520567.2 XM_017018868.1 XM_017018869.1 XM_017018870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
944296 c.1237G>A p.V413I Chr.12: 8841525-8841525 on Build GRCh38
695497 c.1161C>T p.F387F Chr.12: 8841449-8841449 on Build GRCh38
5376180 c.1154G>A p.G385E Chr.12: 8841442-8841442 on Build GRCh38
944295 c.1231A>G p.T411A Chr.12: 8841519-8841519 on Build GRCh38
4956745 c.1197T>A p.A399A Chr.12: 8841485-8841485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61744222 Chr.12: 8841389-8841389 on Build GRCh38
rs765903113 Chr.12: 8841503-8841503 on Build GRCh38
rs776214499 Chr.12: 8841504-8841504 on Build GRCh38
rs750018676 Chr.12: 8841382-8841382 on Build GRCh38
rs752425903 Chr.12: 8841516-8841516 on Build GRCh38
rs763384356 Chr.12: 8841348-8841348 on Build GRCh38
rs765992889 Chr.12: 8841366-8841366 on Build GRCh38
rs185181365 Chr.12: 8841525-8841525 on Build GRCh38
rs756286189 Chr.12: 8841538-8841538 on Build GRCh38
rs368863619 Chr.12: 8841544-8841544 on Build GRCh38
rs769029517 Chr.12: 8841489-8841489 on Build GRCh38
rs766986325 Chr.12: 8841350-8841350 on Build GRCh38
rs760144786 Chr.12: 8841495-8841495 on Build GRCh38
rs75353365 Chr.12: 8841461-8841461 on Build GRCh38
rs375780803 Chr.12: 8841524-8841524 on Build GRCh38
rs770139666 Chr.12: 8841467-8841467 on Build GRCh38
rs921912575 Chr.12: 8841507-8841507 on Build GRCh38
rs920870328 Chr.12: 8841514-8841514 on Build GRCh38
rs760234806 Chr.12: 8841356-8841356 on Build GRCh38
rs767616238 Chr.12: 8841470-8841470 on Build GRCh38
rs760905677 Chr.12: 8841346-8841346 on Build GRCh38
rs749728019 Chr.12: 8841477-8841477 on Build GRCh38
rs61744220 Chr.12: 8841397-8841397 on Build GRCh38
rs267603715 Chr.12: 8841444-8841444 on Build GRCh38
rs372461898 Chr.12: 8841512-8841512 on Build GRCh38
rs763258371 Chr.12: 8841430-8841438 on Build GRCh38
rs374480312 Chr.12: 8841526-8841526 on Build GRCh38
rs777046386 Chr.12: 8841341-8841341 on Build GRCh38
rs74443529 Chr.12: 8841462-8841462 on Build GRCh38
rs749930364 Chr.12: 8841351-8841351 on Build GRCh38
rs371576976 Chr.12: 8841519-8841519 on Build GRCh38
rs774798529 Chr.12: 8841546-8841546 on Build GRCh38
rs78584364 Chr.12: 8841463-8841463 on Build GRCh38
rs776119723 Chr.12: 8841556-8841556 on Build GRCh38
rs377270924 Chr.12: 8841367-8841367 on Build GRCh38
rs773314979 Chr.12: 8841440-8841440 on Build GRCh38
rs757455501 Chr.12: 8841451-8841451 on Build GRCh38
rs746037337 Chr.12: 8841531-8841531 on Build GRCh38
rs370769861 Chr.12: 8841548-8841548 on Build GRCh38
rs774886599 Chr.12: 8841494-8841494 on Build GRCh38
rs369455997 Chr.12: 8841383-8841383 on Build GRCh38
rs375370882 Chr.12: 8841560-8841560 on Build GRCh38
rs1039150694 Chr.12: 8841549-8841549 on Build GRCh38
rs374335176 Chr.12: 8841347-8841347 on Build GRCh38
rs754760292 Chr.12: 8841368-8841368 on Build GRCh38
rs781527794 Chr.12: 8841458-8841458 on Build GRCh38
rs747023059 Chr.12: 8841506-8841506 on Build GRCh38
rs765019682 Chr.12: 8841543-8841543 on Build GRCh38
rs764094825 Chr.12: 8841522-8841522 on Build GRCh38
rs867773372 Chr.12: 8841430-8841430 on Build GRCh38
rs974382868 Chr.12: 8841419-8841419 on Build GRCh38
rs764961744 Chr.12: 8841379-8841379 on Build GRCh38
rs866399982 Chr.12: 8841442-8841442 on Build GRCh38

View Full Product Details